Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47943102-47943347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGA1
Entrez Gene ID
1259
Gene Name
cyclic nucleotide gated channel alpha 1
Gene Aliases
CNCG, CNCG1, CNG-1, CNG1, RCNC1, RCNCa, RCNCalpha, RP49
UniGene
Hs.1323
Gene Chromosome Location
Chr.4: 47935015-48016718 on Build GRCh38
Transcripts
NM_000087.3 NM_001142564.1 XM_005248049.4 XM_011513623.2 XM_017007712.1 NR_125879.1

Gene Symbol
LOC101927157
Entrez Gene ID
101927157
Gene Name
uncharacterized LOC101927157
Gene Aliases
-
UniGene
Hs.134190
Gene Chromosome Location
Chr.4: 47914227-47990737 on Build GRCh38
Transcripts
NM_000087.3 NM_001142564.1 XM_005248049.4 XM_011513623.2 XM_017007712.1 NR_125879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6231403 c.346T>A p.S116T Chr.4: 47943284-47943284 on Build GRCh38
3760650 c.352G>A p.D118N Chr.4: 47943278-47943278 on Build GRCh38
79345 c.347C>G p.S116* Chr.4: 47943283-47943283 on Build GRCh38
6331484 c.597G>A p.K199K Chr.4: 47943240-47943240 on Build GRCh38
1055762 c.417G>T p.K139N Chr.4: 47943213-47943213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867857102 Chr.4: 47943222-47943222 on Build GRCh38
rs769774868 Chr.4: 47943190-47943190 on Build GRCh38
rs745775503 Chr.4: 47943149-47943149 on Build GRCh38
rs748314222 Chr.4: 47943276-47943276 on Build GRCh38
rs147591591 Chr.4: 47943258-47943258 on Build GRCh38
rs766955357 Chr.4: 47943264-47943264 on Build GRCh38
rs766903165 Chr.4: 47943244-47943244 on Build GRCh38
rs889829783 Chr.4: 47943245-47943245 on Build GRCh38
rs763963244 Chr.4: 47943232-47943232 on Build GRCh38
rs1043046270 Chr.4: 47943184-47943184 on Build GRCh38
rs749281255 Chr.4: 47943309-47943309 on Build GRCh38
rs539600817 Chr.4: 47943269-47943269 on Build GRCh38
rs121909600 Chr.4: 47943203-47943203 on Build GRCh38
rs755705235 Chr.4: 47943262-47943262 on Build GRCh38
rs28642966 Chr.4: 47943278-47943278 on Build GRCh38
rs761590177 Chr.4: 47943242-47943242 on Build GRCh38
rs775436707 Chr.4: 47943218-47943218 on Build GRCh38
rs755244560 Chr.4: 47943270-47943270 on Build GRCh38
rs772595474 Chr.4: 47943241-47943241 on Build GRCh38
rs773862362 Chr.4: 47943238-47943238 on Build GRCh38
rs868453867 Chr.4: 47943254-47943254 on Build GRCh38
rs758688361 Chr.4: 47943277-47943277 on Build GRCh38
rs140354960 Chr.4: 47943311-47943311 on Build GRCh38
rs773409199 Chr.4: 47943147-47943147 on Build GRCh38
rs772032069 Chr.4: 47943139-47943139 on Build GRCh38
rs747049717 Chr.4: 47943282-47943282 on Build GRCh38
rs748419423 Chr.4: 47943135-47943135 on Build GRCh38
rs769489990 Chr.4: 47943213-47943214 on Build GRCh38
rs771001514 Chr.4: 47943301-47943301 on Build GRCh38
rs775347192 Chr.4: 47943305-47943305 on Build GRCh38
rs576510245 Chr.4: 47943319-47943319 on Build GRCh38
rs751455936 Chr.4: 47943159-47943159 on Build GRCh38

View Full Product Details