Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127709561-127709786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP157
Entrez Gene ID
286207
Gene Name
cilia and flagella associated protein 157
Gene Aliases
C9orf117
UniGene
Hs.652519
Gene Chromosome Location
Chr.9: 127706261-127716002 on Build GRCh38
Transcripts
NM_001012502.2 XM_006717064.3 XM_011518559.2 XM_017014629.1 XM_017014630.1 XM_017014277.1

Gene Symbol
PTRH1
Entrez Gene ID
138428
Gene Name
peptidyl-tRNA hydrolase 1 homolog
Gene Aliases
C9orf115, PTH1
UniGene
Hs.653265
Gene Chromosome Location
Chr.9: 127695024-127716714 on Build GRCh38
Transcripts
NM_001012502.2 XM_006717064.3 XM_011518559.2 XM_017014629.1 XM_017014630.1 XM_017014277.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7087962 c.381C>A p.R127R Chr.9: 127709641-127709641 on Build GRCh38
1659086 c.390C>T p.F130F Chr.9: 127709650-127709650 on Build GRCh38
1105370 c.357C>T p.F119F Chr.9: 127709617-127709617 on Build GRCh38
1105369 c.307G>A p.E103K Chr.9: 127709567-127709567 on Build GRCh38
1674845 c.373C>T p.Q125* Chr.9: 127709633-127709633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758684799 Chr.9: 127709702-127709702 on Build GRCh38
rs763107425 Chr.9: 127709629-127709629 on Build GRCh38
rs955285048 Chr.9: 127709608-127709608 on Build GRCh38
rs1018231636 Chr.9: 127709603-127709603 on Build GRCh38
rs372752418 Chr.9: 127709645-127709645 on Build GRCh38
rs775593969 Chr.9: 127709627-127709627 on Build GRCh38
rs959527399 Chr.9: 127709714-127709714 on Build GRCh38
rs770747114 Chr.9: 127709599-127709599 on Build GRCh38
rs987769519 Chr.9: 127709612-127709612 on Build GRCh38
rs772829577 Chr.9: 127709595-127709595 on Build GRCh38
rs756501202 Chr.9: 127709659-127709659 on Build GRCh38
rs747616414 Chr.9: 127709586-127709586 on Build GRCh38
rs759381444 Chr.9: 127709617-127709617 on Build GRCh38
rs771592154 Chr.9: 127709594-127709594 on Build GRCh38
rs369180901 Chr.9: 127709640-127709640 on Build GRCh38
rs754390603 Chr.9: 127709674-127709674 on Build GRCh38
rs761954321 Chr.9: 127709731-127709731 on Build GRCh38
rs764714243 Chr.9: 127709696-127709697 on Build GRCh38
rs775540886 Chr.9: 127709718-127709718 on Build GRCh38
rs763046389 Chr.9: 127709727-127709727 on Build GRCh38
rs377409854 Chr.9: 127709741-127709741 on Build GRCh38
rs750685095 Chr.9: 127709652-127709652 on Build GRCh38
rs765346649 Chr.9: 127709636-127709636 on Build GRCh38
rs201158784 Chr.9: 127709685-127709685 on Build GRCh38
rs963690829 Chr.9: 127709623-127709623 on Build GRCh38
rs950310480 Chr.9: 127709736-127709736 on Build GRCh38
rs769765087 Chr.9: 127709713-127709713 on Build GRCh38
rs375903663 Chr.9: 127709676-127709676 on Build GRCh38
rs746707316 Chr.9: 127709597-127709597 on Build GRCh38
rs765229107 Chr.9: 127709618-127709618 on Build GRCh38
rs767592297 Chr.9: 127709733-127709733 on Build GRCh38
rs776233369 Chr.9: 127709606-127709606 on Build GRCh38
rs115025333 Chr.9: 127709730-127709730 on Build GRCh38
rs1047012595 Chr.9: 127709752-127709752 on Build GRCh38
rs200871397 Chr.9: 127709704-127709704 on Build GRCh38
rs764305353 Chr.9: 127709639-127709639 on Build GRCh38
rs780542156 Chr.9: 127709581-127709581 on Build GRCh38
rs768839920 Chr.9: 127709729-127709729 on Build GRCh38
rs746543146 Chr.9: 127709699-127709699 on Build GRCh38
rs745604179 Chr.9: 127709710-127709710 on Build GRCh38
rs757476136 Chr.9: 127709643-127709643 on Build GRCh38
rs780489169 Chr.9: 127709664-127709664 on Build GRCh38

View Full Product Details