Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36319497-36319750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf222
Entrez Gene ID
389384
Gene Name
chromosome 6 open reading frame 222
Gene Aliases
-
UniGene
Hs.162104
Gene Chromosome Location
Chr.6: 36315757-36337015 on Build GRCh38
Transcripts
NM_001010903.4 XM_011514596.2 XM_011514597.2 XM_011514598.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1444094 c.1702delT p.Y568fs*12 Chr.6: 36319577-36319577 on Build GRCh38
5902296 c.1769C>T p.S590F Chr.6: 36319510-36319510 on Build GRCh38
3394152 c.1702T>C p.Y568H Chr.6: 36319577-36319577 on Build GRCh38
6361154 c.1773G>T p.K591N Chr.6: 36319506-36319506 on Build GRCh38
3626225 c.1705G>A p.E569K Chr.6: 36319574-36319574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574214842 Chr.6: 36319690-36319690 on Build GRCh38
rs375210086 Chr.6: 36319603-36319603 on Build GRCh38
rs774087270 Chr.6: 36319536-36319536 on Build GRCh38
rs767403088 Chr.6: 36319547-36319547 on Build GRCh38
rs917027288 Chr.6: 36319683-36319683 on Build GRCh38
rs752100110 Chr.6: 36319658-36319658 on Build GRCh38
rs138094812 Chr.6: 36319574-36319574 on Build GRCh38
rs759873742 Chr.6: 36319549-36319549 on Build GRCh38
rs571853090 Chr.6: 36319663-36319663 on Build GRCh38
rs776711424 Chr.6: 36319646-36319646 on Build GRCh38
rs772098689 Chr.6: 36319621-36319621 on Build GRCh38
rs879714763 Chr.6: 36319599-36319599 on Build GRCh38
rs758345455 Chr.6: 36319567-36319567 on Build GRCh38
rs373450017 Chr.6: 36319532-36319532 on Build GRCh38
rs761762985 Chr.6: 36319546-36319546 on Build GRCh38
rs755783603 Chr.6: 36319577-36319577 on Build GRCh38
rs142695827 Chr.6: 36319575-36319575 on Build GRCh38
rs187458829 Chr.6: 36319570-36319570 on Build GRCh38
rs768336137 Chr.6: 36319535-36319535 on Build GRCh38
rs369990047 Chr.6: 36319566-36319566 on Build GRCh38
rs757792547 Chr.6: 36319660-36319660 on Build GRCh38
rs80279351 Chr.6: 36319692-36319692 on Build GRCh38
rs746290197 Chr.6: 36319519-36319526 on Build GRCh38
rs201478093 Chr.6: 36319556-36319556 on Build GRCh38
rs984795124 Chr.6: 36319698-36319698 on Build GRCh38
rs967530559 Chr.6: 36319701-36319701 on Build GRCh38
rs750912939 Chr.6: 36319719-36319719 on Build GRCh38
rs743852 Chr.6: 36319535-36319535 on Build GRCh38
rs553438211 Chr.6: 36319661-36319661 on Build GRCh38
rs769333185 Chr.6: 36319531-36319531 on Build GRCh38
rs895745702 Chr.6: 36319587-36319587 on Build GRCh38
rs190406294 Chr.6: 36319614-36319614 on Build GRCh38
rs773166385 Chr.6: 36319630-36319630 on Build GRCh38
rs538483483 Chr.6: 36319655-36319655 on Build GRCh38
rs764060346 Chr.6: 36319656-36319656 on Build GRCh38
rs535797169 Chr.6: 36319664-36319664 on Build GRCh38
rs768042350 Chr.6: 36319604-36319604 on Build GRCh38
rs779512450 Chr.6: 36319584-36319584 on Build GRCh38
rs778475187 Chr.6: 36319571-36319572 on Build GRCh38
rs747868798 Chr.6: 36319576-36319577 on Build GRCh38
rs760491286 Chr.6: 36319640-36319640 on Build GRCh38
rs554556810 Chr.6: 36319669-36319669 on Build GRCh38
rs373354401 Chr.6: 36319579-36319579 on Build GRCh38
rs763151511 Chr.6: 36319647-36319647 on Build GRCh38

View Full Product Details