Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7503962-7504227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf45
Entrez Gene ID
374877
Gene Name
chromosome 19 open reading frame 45
Gene Aliases
-
UniGene
Hs.631862
Gene Chromosome Location
Chr.19: 7497559-7508457 on Build GRCh38
Transcripts
NM_198534.2 XM_005272473.3 XM_011527993.1 XM_011527995.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6017168 c.578-5C>T p.? Chr.19: 7504116-7504116 on Build GRCh38
148471 c.620C>G p.P207R Chr.19: 7504205-7504205 on Build GRCh38
5517713 c.605A>G p.Y202C Chr.19: 7504148-7504148 on Build GRCh38
6038105 c.628T>C p.Y210H Chr.19: 7504171-7504171 on Build GRCh38
4553044 c.537G>A p.G179G Chr.19: 7504122-7504122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915562120 Chr.19: 7504081-7504082 on Build GRCh38
rs770288362 Chr.19: 7504100-7504100 on Build GRCh38
rs1026923282 Chr.19: 7504012-7504012 on Build GRCh38
rs17256868 Chr.19: 7503993-7503993 on Build GRCh38
rs552737442 Chr.19: 7504151-7504151 on Build GRCh38
rs773914284 Chr.19: 7504101-7504101 on Build GRCh38
rs959893852 Chr.19: 7504188-7504188 on Build GRCh38
rs775328015 Chr.19: 7504105-7504105 on Build GRCh38
rs781463689 Chr.19: 7504079-7504079 on Build GRCh38
rs751658567 Chr.19: 7504134-7504134 on Build GRCh38
rs201900726 Chr.19: 7504152-7504152 on Build GRCh38
rs557310468 Chr.19: 7504076-7504076 on Build GRCh38
rs113357637 Chr.19: 7504095-7504095 on Build GRCh38
rs796376367 Chr.19: 7504099-7504100 on Build GRCh38
rs997628454 Chr.19: 7504031-7504031 on Build GRCh38
rs188990556 Chr.19: 7504111-7504111 on Build GRCh38
rs749688912 Chr.19: 7504167-7504167 on Build GRCh38
rs770193911 Chr.19: 7504097-7504097 on Build GRCh38
rs551538195 Chr.19: 7504021-7504021 on Build GRCh38
rs891804971 Chr.19: 7504057-7504057 on Build GRCh38
rs771743139 Chr.19: 7504191-7504191 on Build GRCh38
rs771464530 Chr.19: 7504174-7504174 on Build GRCh38
rs778212136 Chr.19: 7504161-7504161 on Build GRCh38
rs138148015 Chr.19: 7504205-7504205 on Build GRCh38
rs184442770 Chr.19: 7504098-7504098 on Build GRCh38
rs755109389 Chr.19: 7504140-7504140 on Build GRCh38
rs745345973 Chr.19: 7504180-7504180 on Build GRCh38
rs761825589 Chr.19: 7504127-7504127 on Build GRCh38
rs192488102 Chr.19: 7504000-7504000 on Build GRCh38
rs150394805 Chr.19: 7504039-7504039 on Build GRCh38
rs1010208747 Chr.19: 7504093-7504093 on Build GRCh38
rs760513547 Chr.19: 7504110-7504110 on Build GRCh38
rs200940379 Chr.19: 7504198-7504198 on Build GRCh38
rs780208505 Chr.19: 7504070-7504070 on Build GRCh38
rs755437535 Chr.19: 7504099-7504100 on Build GRCh38
rs113706007 Chr.19: 7504080-7504081 on Build GRCh38
rs371188525 Chr.19: 7504104-7504104 on Build GRCh38
rs60571341 Chr.19: 7504094-7504095 on Build GRCh38
rs759035622 Chr.19: 7504103-7504103 on Build GRCh38
rs571381920 Chr.19: 7504029-7504029 on Build GRCh38
rs753672467 Chr.19: 7504120-7504120 on Build GRCh38
rs762795386 Chr.19: 7504093-7504094 on Build GRCh38
rs867054622 Chr.19: 7504080-7504080 on Build GRCh38
rs749834281 Chr.19: 7504099-7504099 on Build GRCh38
rs766306975 Chr.19: 7504132-7504132 on Build GRCh38
rs754105587 Chr.19: 7504097-7504100 on Build GRCh38

View Full Product Details