Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42865247-42865484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACKR2
Entrez Gene ID
1238
Gene Name
atypical chemokine receptor 2
Gene Aliases
CCBP2, CCR10, CCR9, CMKBR9, D6, hD6
UniGene
Hs.146346
Gene Chromosome Location
Chr.3: 42809472-42867283 on Build GRCh38
Transcripts
NM_001296.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
583839 c.923C>T p.S308F Chr.3: 42865425-42865425 on Build GRCh38
95796 c.876C>T p.Y292Y Chr.3: 42865378-42865378 on Build GRCh38
5746112 c.946A>G p.S316G Chr.3: 42865448-42865448 on Build GRCh38
4117662 c.875A>G p.Y292C Chr.3: 42865377-42865377 on Build GRCh38
1044795 c.901G>A p.A301T Chr.3: 42865403-42865403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541554327 Chr.3: 42865320-42865320 on Build GRCh38
rs771353336 Chr.3: 42865301-42865301 on Build GRCh38
rs767623264 Chr.3: 42865348-42865348 on Build GRCh38
rs771149260 Chr.3: 42865351-42865351 on Build GRCh38
rs142416155 Chr.3: 42865379-42865379 on Build GRCh38
rs752768615 Chr.3: 42865349-42865349 on Build GRCh38
rs377711584 Chr.3: 42865279-42865279 on Build GRCh38
rs370078092 Chr.3: 42865454-42865454 on Build GRCh38
rs768208057 Chr.3: 42865383-42865383 on Build GRCh38
rs139094169 Chr.3: 42865286-42865286 on Build GRCh38
rs200987829 Chr.3: 42865402-42865402 on Build GRCh38
rs765210048 Chr.3: 42865275-42865275 on Build GRCh38
rs752964827 Chr.3: 42865288-42865288 on Build GRCh38
rs778062440 Chr.3: 42865293-42865293 on Build GRCh38
rs761612800 Chr.3: 42865339-42865339 on Build GRCh38
rs776347264 Chr.3: 42865330-42865330 on Build GRCh38
rs765678623 Chr.3: 42865448-42865448 on Build GRCh38
rs376634810 Chr.3: 42865360-42865360 on Build GRCh38
rs35951725 Chr.3: 42865327-42865328 on Build GRCh38
rs138890782 Chr.3: 42865461-42865461 on Build GRCh38
rs6779520 Chr.3: 42865433-42865433 on Build GRCh38
rs371264370 Chr.3: 42865419-42865419 on Build GRCh38
rs762854358 Chr.3: 42865346-42865346 on Build GRCh38
rs189203964 Chr.3: 42865451-42865451 on Build GRCh38
rs767125330 Chr.3: 42865438-42865439 on Build GRCh38
rs368719352 Chr.3: 42865271-42865271 on Build GRCh38
rs764251333 Chr.3: 42865435-42865435 on Build GRCh38
rs760448133 Chr.3: 42865323-42865323 on Build GRCh38
rs774968799 Chr.3: 42865393-42865394 on Build GRCh38
rs746429853 Chr.3: 42865375-42865375 on Build GRCh38
rs754045457 Chr.3: 42865446-42865446 on Build GRCh38
rs1056643270 Chr.3: 42865324-42865324 on Build GRCh38
rs745405456 Chr.3: 42865319-42865319 on Build GRCh38
rs757599752 Chr.3: 42865369-42865369 on Build GRCh38
rs765093153 Chr.3: 42865340-42865340 on Build GRCh38
rs202107471 Chr.3: 42865291-42865291 on Build GRCh38
rs776281860 Chr.3: 42865387-42865387 on Build GRCh38
rs561300072 Chr.3: 42865345-42865345 on Build GRCh38
rs547304854 Chr.3: 42865378-42865378 on Build GRCh38
rs955213024 Chr.3: 42865429-42865429 on Build GRCh38
rs748837237 Chr.3: 42865300-42865300 on Build GRCh38
rs199831111 Chr.3: 42865457-42865457 on Build GRCh38
rs1026585413 Chr.3: 42865351-42865351 on Build GRCh38
rs772959627 Chr.3: 42865408-42865408 on Build GRCh38
rs199910446 Chr.3: 42865427-42865427 on Build GRCh38
rs144905810 Chr.3: 42865372-42865372 on Build GRCh38
rs755009181 Chr.3: 42865280-42865280 on Build GRCh38
rs184802180 Chr.3: 42865449-42865449 on Build GRCh38
rs1043623168 Chr.3: 42865294-42865294 on Build GRCh38
rs896689766 Chr.3: 42865342-42865342 on Build GRCh38
rs759931729 Chr.3: 42865435-42865436 on Build GRCh38
rs777989602 Chr.3: 42865357-42865357 on Build GRCh38
rs371241795 Chr.3: 42865302-42865302 on Build GRCh38
rs754472756 Chr.3: 42865460-42865460 on Build GRCh38
rs747916833 Chr.3: 42865391-42865391 on Build GRCh38
rs762945000 Chr.3: 42865426-42865426 on Build GRCh38
rs868771798 Chr.3: 42865425-42865425 on Build GRCh38
rs373940627 Chr.3: 42865353-42865353 on Build GRCh38

View Full Product Details