Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129421328-129421601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB12
Entrez Gene ID
90288
Gene Name
EF-hand calcium binding domain 12
Gene Aliases
C3orf25
UniGene
Hs.652347
Gene Chromosome Location
Chr.3: 129401321-129428651 on Build GRCh38
Transcripts
NM_207307.2 XM_011513293.2 XM_011513294.2 XM_011513295.2 XM_011513297.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667699 c.433G>A p.E145K Chr.3: 129421420-129421420 on Build GRCh38
6272085 c.448C>A p.Q150K Chr.3: 129421405-129421405 on Build GRCh38
4761251 c.470C>T p.T157I Chr.3: 129421383-129421383 on Build GRCh38
5903456 c.353C>T p.S118F Chr.3: 129421500-129421500 on Build GRCh38
3846165 c.401C>A p.P134H Chr.3: 129421452-129421452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755755799 Chr.3: 129421573-129421573 on Build GRCh38
rs551740547 Chr.3: 129421563-129421563 on Build GRCh38
rs775576129 Chr.3: 129421528-129421528 on Build GRCh38
rs781460468 Chr.3: 129421574-129421574 on Build GRCh38
rs747468883 Chr.3: 129421515-129421515 on Build GRCh38
rs1005940039 Chr.3: 129421387-129421387 on Build GRCh38
rs773745745 Chr.3: 129421403-129421403 on Build GRCh38
rs372347992 Chr.3: 129421465-129421465 on Build GRCh38
rs140620659 Chr.3: 129421358-129421358 on Build GRCh38
rs533576967 Chr.3: 129421381-129421381 on Build GRCh38
rs764032358 Chr.3: 129421544-129421544 on Build GRCh38
rs368934436 Chr.3: 129421455-129421455 on Build GRCh38
rs914599068 Chr.3: 129421577-129421577 on Build GRCh38
rs201014037 Chr.3: 129421516-129421516 on Build GRCh38
rs759724226 Chr.3: 129421431-129421431 on Build GRCh38
rs769039790 Chr.3: 129421363-129421363 on Build GRCh38
rs751351089 Chr.3: 129421559-129421559 on Build GRCh38
rs773657576 Chr.3: 129421510-129421510 on Build GRCh38
rs763834383 Chr.3: 129421453-129421453 on Build GRCh38
rs758352542 Chr.3: 129421443-129421443 on Build GRCh38
rs772518314 Chr.3: 129421393-129421393 on Build GRCh38
rs769861768 Chr.3: 129421526-129421526 on Build GRCh38
rs777981572 Chr.3: 129421405-129421405 on Build GRCh38
rs189462288 Chr.3: 129421410-129421410 on Build GRCh38
rs772714202 Chr.3: 129421502-129421502 on Build GRCh38
rs1055928401 Chr.3: 129421356-129421356 on Build GRCh38
rs762809624 Chr.3: 129421542-129421542 on Build GRCh38
rs779428255 Chr.3: 129421576-129421576 on Build GRCh38
rs201855762 Chr.3: 129421567-129421567 on Build GRCh38
rs765050485 Chr.3: 129421432-129421432 on Build GRCh38
rs779735030 Chr.3: 129421490-129421490 on Build GRCh38
rs547269424 Chr.3: 129421429-129421430 on Build GRCh38
rs752678319 Chr.3: 129421442-129421442 on Build GRCh38
rs760916534 Chr.3: 129421406-129421406 on Build GRCh38
rs1006880256 Chr.3: 129421547-129421547 on Build GRCh38
rs751359158 Chr.3: 129421454-129421454 on Build GRCh38
rs776556788 Chr.3: 129421420-129421420 on Build GRCh38
rs776949033 Chr.3: 129421523-129421523 on Build GRCh38
rs143690542 Chr.3: 129421524-129421524 on Build GRCh38
rs371476230 Chr.3: 129421569-129421569 on Build GRCh38
rs755226807 Chr.3: 129421460-129421460 on Build GRCh38
rs573378171 Chr.3: 129421388-129421388 on Build GRCh38
rs755700164 Chr.3: 129421482-129421482 on Build GRCh38
rs780502219 Chr.3: 129421350-129421350 on Build GRCh38

View Full Product Details