Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50614154-50614417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATL1
Entrez Gene ID
51062
Gene Name
atlastin GTPase 1
Gene Aliases
AD-FSP, FSP1, GBP3, HSN1D, SPG3, SPG3A, atlastin1
UniGene
Hs.584905
Gene Chromosome Location
Chr.14: 50533082-50633068 on Build GRCh38
Transcripts
NM_001127713.1 NM_015915.4 NM_181598.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6358665 c.743A>T p.E248V Chr.14: 50614392-50614392 on Build GRCh38
2198599 c.756C>T p.N252N Chr.14: 50614405-50614405 on Build GRCh38
1748872 c.764A>C p.K255T Chr.14: 50614413-50614413 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs534627563 Chr.14: 50614232-50614232 on Build GRCh38
rs533376430 Chr.14: 50614364-50614364 on Build GRCh38
rs190301749 Chr.14: 50614191-50614191 on Build GRCh38
rs111677408 Chr.14: 50614274-50614274 on Build GRCh38
rs557991405 Chr.14: 50614258-50614258 on Build GRCh38
rs916872404 Chr.14: 50614361-50614361 on Build GRCh38
rs182278371 Chr.14: 50614301-50614301 on Build GRCh38
rs996060447 Chr.14: 50614201-50614201 on Build GRCh38
rs147727059 Chr.14: 50614189-50614189 on Build GRCh38
rs747797630 Chr.14: 50614360-50614360 on Build GRCh38
rs142624440 Chr.14: 50614342-50614342 on Build GRCh38
rs758965544 Chr.14: 50614337-50614337 on Build GRCh38
rs1030451134 Chr.14: 50614352-50614352 on Build GRCh38
rs780733244 Chr.14: 50614354-50614354 on Build GRCh38
rs1028916511 Chr.14: 50614216-50614216 on Build GRCh38
rs539233428 Chr.14: 50614298-50614298 on Build GRCh38
rs566041285 Chr.14: 50614182-50614182 on Build GRCh38
rs370498300 Chr.14: 50614236-50614236 on Build GRCh38
rs748661789 Chr.14: 50614200-50614200 on Build GRCh38
rs543016813 Chr.14: 50614228-50614228 on Build GRCh38
rs748802949 Chr.14: 50614390-50614390 on Build GRCh38
rs755348062 Chr.14: 50614346-50614346 on Build GRCh38
rs577600870 Chr.14: 50614220-50614220 on Build GRCh38
rs1055952048 Chr.14: 50614283-50614283 on Build GRCh38
rs897571836 Chr.14: 50614302-50614302 on Build GRCh38
rs901521411 Chr.14: 50614197-50614197 on Build GRCh38
rs772913174 Chr.14: 50614373-50614373 on Build GRCh38
rs770124315 Chr.14: 50614259-50614259 on Build GRCh38
rs942215891 Chr.14: 50614300-50614300 on Build GRCh38
rs747415606 Chr.14: 50614344-50614344 on Build GRCh38

View Full Product Details