Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110152317-110152546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A1
Entrez Gene ID
1282
Gene Name
collagen type IV alpha 1 chain
Gene Aliases
BSVD, RATOR
UniGene
Hs.17441
Gene Chromosome Location
Chr.13: 110148958-110307157 on Build GRCh38
Transcripts
NM_001845.5 XM_011521048.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
384610 c.4805G>T p.G1602V Chr.13: 110152457-110152457 on Build GRCh38
5362153 c.3785G>A p.R1262H Chr.13: 110152406-110152406 on Build GRCh38
4496870 c.4849C>T p.H1617Y Chr.13: 110152413-110152413 on Build GRCh38
4045911 c.4796C>T p.A1599V Chr.13: 110152466-110152466 on Build GRCh38
3872820 c.4808C>T p.S1603F Chr.13: 110152454-110152454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565252213 Chr.13: 110152526-110152526 on Build GRCh38
rs771443426 Chr.13: 110152497-110152497 on Build GRCh38
rs148750080 Chr.13: 110152387-110152387 on Build GRCh38
rs672601348 Chr.13: 110152381-110152381 on Build GRCh38
rs915529489 Chr.13: 110152432-110152432 on Build GRCh38
rs746538445 Chr.13: 110152518-110152518 on Build GRCh38
rs984568560 Chr.13: 110152378-110152378 on Build GRCh38
rs778082936 Chr.13: 110152493-110152493 on Build GRCh38
rs375486886 Chr.13: 110152520-110152520 on Build GRCh38
rs765084929 Chr.13: 110152463-110152463 on Build GRCh38
rs1040826004 Chr.13: 110152521-110152521 on Build GRCh38
rs753054366 Chr.13: 110152477-110152477 on Build GRCh38
rs779680003 Chr.13: 110152498-110152498 on Build GRCh38
rs766472965 Chr.13: 110152466-110152466 on Build GRCh38
rs765269741 Chr.13: 110152386-110152386 on Build GRCh38
rs144201848 Chr.13: 110152380-110152380 on Build GRCh38
rs749824018 Chr.13: 110152494-110152494 on Build GRCh38
rs760487355 Chr.13: 110152363-110152363 on Build GRCh38
rs146638269 Chr.13: 110152465-110152465 on Build GRCh38
rs751674518 Chr.13: 110152467-110152467 on Build GRCh38
rs758573884 Chr.13: 110152390-110152390 on Build GRCh38
rs889464899 Chr.13: 110152484-110152484 on Build GRCh38
rs755291402 Chr.13: 110152407-110152407 on Build GRCh38
rs190097900 Chr.13: 110152459-110152459 on Build GRCh38
rs781449099 Chr.13: 110152471-110152471 on Build GRCh38
rs748558670 Chr.13: 110152411-110152411 on Build GRCh38
rs140661222 Chr.13: 110152438-110152438 on Build GRCh38
rs776425169 Chr.13: 110152522-110152522 on Build GRCh38
rs745307916 Chr.13: 110152428-110152428 on Build GRCh38
rs755166798 Chr.13: 110152470-110152470 on Build GRCh38
rs973027770 Chr.13: 110152342-110152342 on Build GRCh38
rs747250117 Chr.13: 110152406-110152406 on Build GRCh38
rs200168799 Chr.13: 110152527-110152527 on Build GRCh38
rs146855373 Chr.13: 110152420-110152420 on Build GRCh38
rs650724 Chr.13: 110152462-110152462 on Build GRCh38
rs542803991 Chr.13: 110152375-110152375 on Build GRCh38
rs756504354 Chr.13: 110152486-110152486 on Build GRCh38
rs987149771 Chr.13: 110152423-110152423 on Build GRCh38
rs778511113 Chr.13: 110152510-110152511 on Build GRCh38
rs201012509 Chr.13: 110152355-110152355 on Build GRCh38
rs771624014 Chr.13: 110152429-110152429 on Build GRCh38
rs149495996 Chr.13: 110152362-110152362 on Build GRCh38
rs780076830 Chr.13: 110152394-110152394 on Build GRCh38
rs781502396 Chr.13: 110152410-110152410 on Build GRCh38
rs142326550 Chr.13: 110152417-110152417 on Build GRCh38
rs775233741 Chr.13: 110152430-110152430 on Build GRCh38
rs1055738385 Chr.13: 110152489-110152489 on Build GRCh38
rs753360259 Chr.13: 110152344-110152344 on Build GRCh38

View Full Product Details