Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76331145-76331418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC158
Entrez Gene ID
339965
Gene Name
coiled-coil domain containing 158
Gene Aliases
-
UniGene
Hs.529680
Gene Chromosome Location
Chr.4: 76313039-76421920 on Build GRCh38
Transcripts
NM_001042784.1 XM_005262974.4 XM_006714204.3 XM_011531908.2 XM_011531909.2 XM_011531910.2 XM_011531911.1 XM_011531912.2 XM_011531913.1 XM_011531914.2 XM_011531915.2 XM_011531916.2 XM_011531917.1 XM_017008160.1 XM_017008161.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2151068 c.2883G>A p.S961S Chr.4: 76331391-76331391 on Build GRCh38
6210371 c.2871-7C>T p.? Chr.4: 76331410-76331410 on Build GRCh38
4126015 c.2873G>A p.S958N Chr.4: 76331401-76331401 on Build GRCh38
1619084 c.2882C>T p.S961L Chr.4: 76331392-76331392 on Build GRCh38
6478679 c.2899G>A p.E967K Chr.4: 76331375-76331375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532287424 Chr.4: 76331207-76331207 on Build GRCh38
rs192439362 Chr.4: 76331309-76331309 on Build GRCh38
rs746990745 Chr.4: 76331397-76331397 on Build GRCh38
rs560337298 Chr.4: 76331190-76331190 on Build GRCh38
rs774450491 Chr.4: 76331183-76331183 on Build GRCh38
rs899945866 Chr.4: 76331284-76331284 on Build GRCh38
rs943902827 Chr.4: 76331237-76331237 on Build GRCh38
rs964578419 Chr.4: 76331372-76331372 on Build GRCh38
rs894340978 Chr.4: 76331210-76331210 on Build GRCh38
rs570121273 Chr.4: 76331330-76331330 on Build GRCh38
rs756577328 Chr.4: 76331354-76331354 on Build GRCh38
rs557849729 Chr.4: 76331326-76331326 on Build GRCh38
rs188161731 Chr.4: 76331208-76331208 on Build GRCh38
rs772001135 Chr.4: 76331312-76331312 on Build GRCh38
rs907089392 Chr.4: 76331253-76331253 on Build GRCh38
rs778561830 Chr.4: 76331383-76331383 on Build GRCh38
rs562772829 Chr.4: 76331266-76331266 on Build GRCh38
rs1046733119 Chr.4: 76331187-76331187 on Build GRCh38
rs754208793 Chr.4: 76331358-76331358 on Build GRCh38
rs201892546 Chr.4: 76331329-76331329 on Build GRCh38
rs757901286 Chr.4: 76331297-76331297 on Build GRCh38
rs763138533 Chr.4: 76331328-76331328 on Build GRCh38
rs376541300 Chr.4: 76331368-76331368 on Build GRCh38
rs754644725 Chr.4: 76331367-76331367 on Build GRCh38
rs771522809 Chr.4: 76331392-76331392 on Build GRCh38
rs780320632 Chr.4: 76331356-76331356 on Build GRCh38
rs1046076139 Chr.4: 76331244-76331244 on Build GRCh38
rs770595090 Chr.4: 76331324-76331324 on Build GRCh38
rs58968074 Chr.4: 76331294-76331294 on Build GRCh38
rs1012732679 Chr.4: 76331229-76331229 on Build GRCh38
rs777839658 Chr.4: 76331345-76331348 on Build GRCh38
rs768149486 Chr.4: 76331339-76331339 on Build GRCh38
rs908361194 Chr.4: 76331212-76331212 on Build GRCh38
rs980477661 Chr.4: 76331182-76331182 on Build GRCh38
rs373587869 Chr.4: 76331349-76331349 on Build GRCh38
rs760486186 Chr.4: 76331314-76331314 on Build GRCh38
rs79867003 Chr.4: 76331303-76331303 on Build GRCh38
rs771944838 Chr.4: 76331299-76331299 on Build GRCh38
rs374627865 Chr.4: 76331302-76331302 on Build GRCh38
rs761914152 Chr.4: 76331337-76331337 on Build GRCh38
rs533975151 Chr.4: 76331305-76331305 on Build GRCh38
rs1024703020 Chr.4: 76331395-76331395 on Build GRCh38
rs376345467 Chr.4: 76331391-76331391 on Build GRCh38
rs922855809 Chr.4: 76331177-76331177 on Build GRCh38
rs919802504 Chr.4: 76331184-76331184 on Build GRCh38
rs773078008 Chr.4: 76331313-76331313 on Build GRCh38
rs777721428 Chr.4: 76331393-76331393 on Build GRCh38

View Full Product Details