Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40166690-40166933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC3
Entrez Gene ID
22794
Gene Name
cancer susceptibility candidate 3
Gene Aliases
BTZ, MLN51
UniGene
Hs.743287
Gene Chromosome Location
Chr.17: 40140254-40172178 on Build GRCh38
Transcripts
NM_007359.4 XM_005257163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6283035 c.1531G>A p.E511K Chr.17: 40166856-40166856 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781070268 Chr.17: 40166836-40166836 on Build GRCh38
rs753190696 Chr.17: 40166794-40166794 on Build GRCh38
rs918577365 Chr.17: 40166873-40166873 on Build GRCh38
rs767635283 Chr.17: 40166754-40166754 on Build GRCh38
rs758494101 Chr.17: 40166755-40166755 on Build GRCh38
rs753240013 Chr.17: 40166896-40166896 on Build GRCh38
rs200973321 Chr.17: 40166880-40166880 on Build GRCh38
rs375416237 Chr.17: 40166779-40166779 on Build GRCh38
rs1029259834 Chr.17: 40166727-40166727 on Build GRCh38
rs34645043 Chr.17: 40166818-40166819 on Build GRCh38
rs150381393 Chr.17: 40166800-40166800 on Build GRCh38
rs773292819 Chr.17: 40166872-40166872 on Build GRCh38
rs74458287 Chr.17: 40166898-40166898 on Build GRCh38
rs953207853 Chr.17: 40166745-40166745 on Build GRCh38
rs746642611 Chr.17: 40166814-40166814 on Build GRCh38
rs760954302 Chr.17: 40166758-40166758 on Build GRCh38
rs972761752 Chr.17: 40166865-40166865 on Build GRCh38
rs774258838 Chr.17: 40166749-40166749 on Build GRCh38
rs938525047 Chr.17: 40166738-40166738 on Build GRCh38
rs866265516 Chr.17: 40166802-40166802 on Build GRCh38
rs370392697 Chr.17: 40166887-40166887 on Build GRCh38
rs1043682418 Chr.17: 40166895-40166895 on Build GRCh38
rs770725087 Chr.17: 40166823-40166823 on Build GRCh38
rs779495613 Chr.17: 40166786-40166786 on Build GRCh38
rs762839620 Chr.17: 40166854-40166854 on Build GRCh38
rs1056691094 Chr.17: 40166883-40166883 on Build GRCh38
rs200411531 Chr.17: 40166900-40166900 on Build GRCh38
rs962783767 Chr.17: 40166809-40166809 on Build GRCh38
rs745709047 Chr.17: 40166846-40166846 on Build GRCh38
rs369066110 Chr.17: 40166847-40166847 on Build GRCh38
rs772135625 Chr.17: 40166859-40166859 on Build GRCh38
rs777488293 Chr.17: 40166805-40166805 on Build GRCh38
rs780156915 Chr.17: 40166848-40166848 on Build GRCh38
rs754233567 Chr.17: 40166763-40166763 on Build GRCh38
rs372414129 Chr.17: 40166875-40166875 on Build GRCh38
rs759920358 Chr.17: 40166886-40166886 on Build GRCh38
rs1036099030 Chr.17: 40166721-40166721 on Build GRCh38
rs907065828 Chr.17: 40166736-40166736 on Build GRCh38
rs760663399 Chr.17: 40166895-40166909 on Build GRCh38
rs759028084 Chr.17: 40166897-40166897 on Build GRCh38
rs754100975 Chr.17: 40166885-40166885 on Build GRCh38
rs766672675 Chr.17: 40166759-40166759 on Build GRCh38

View Full Product Details