Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70179459-70179718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf42
Entrez Gene ID
54980
Gene Name
chromosome 2 open reading frame 42
Gene Aliases
-
UniGene
Hs.720049
Gene Chromosome Location
Chr.2: 70149885-70191071 on Build GRCh38
Transcripts
NM_017880.1 XM_005264389.2 XM_005264390.2 XM_005264391.2 XM_011532937.2 XM_017004398.1 XM_017004399.1 XM_017004400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3730652 c.824-3delT p.? Chr.2: 70179645-70179645 on Build GRCh38
3839897 c.865G>C p.E289Q Chr.2: 70179601-70179601 on Build GRCh38
6615835 c.839T>G p.I280S Chr.2: 70179627-70179627 on Build GRCh38
5862765 c.928G>A p.V310I Chr.2: 70179538-70179538 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372665510 Chr.2: 70179608-70179608 on Build GRCh38
rs145895072 Chr.2: 70179621-70179621 on Build GRCh38
rs756194826 Chr.2: 70179638-70179638 on Build GRCh38
rs749583797 Chr.2: 70179649-70179649 on Build GRCh38
rs149251706 Chr.2: 70179631-70179631 on Build GRCh38
rs751863780 Chr.2: 70179617-70179617 on Build GRCh38
rs531731689 Chr.2: 70179521-70179521 on Build GRCh38
rs764573773 Chr.2: 70179611-70179611 on Build GRCh38
rs772871165 Chr.2: 70179569-70179569 on Build GRCh38
rs745733890 Chr.2: 70179676-70179676 on Build GRCh38
rs776739680 Chr.2: 70179673-70179673 on Build GRCh38
rs368286851 Chr.2: 70179578-70179578 on Build GRCh38
rs764799889 Chr.2: 70179595-70179595 on Build GRCh38
rs141350086 Chr.2: 70179586-70179586 on Build GRCh38
rs988984584 Chr.2: 70179495-70179495 on Build GRCh38
rs745928657 Chr.2: 70179513-70179513 on Build GRCh38
rs775208093 Chr.2: 70179605-70179605 on Build GRCh38
rs770958209 Chr.2: 70179583-70179583 on Build GRCh38
rs770800376 Chr.2: 70179672-70179672 on Build GRCh38
rs756703212 Chr.2: 70179651-70179653 on Build GRCh38
rs896954295 Chr.2: 70179674-70179674 on Build GRCh38
rs768036808 Chr.2: 70179625-70179625 on Build GRCh38
rs369768729 Chr.2: 70179648-70179648 on Build GRCh38
rs780290738 Chr.2: 70179532-70179532 on Build GRCh38
rs371516761 Chr.2: 70179487-70179487 on Build GRCh38
rs771915914 Chr.2: 70179543-70179543 on Build GRCh38
rs759057388 Chr.2: 70179680-70179680 on Build GRCh38
rs754515766 Chr.2: 70179487-70179487 on Build GRCh38
rs938291045 Chr.2: 70179594-70179594 on Build GRCh38
rs147618030 Chr.2: 70179589-70179589 on Build GRCh38
rs925537072 Chr.2: 70179592-70179592 on Build GRCh38
rs769438679 Chr.2: 70179684-70179684 on Build GRCh38
rs548640299 Chr.2: 70179537-70179537 on Build GRCh38
rs373914151 Chr.2: 70179657-70179657 on Build GRCh38
rs781676869 Chr.2: 70179644-70179645 on Build GRCh38
rs781314272 Chr.2: 70179502-70179502 on Build GRCh38
rs748618504 Chr.2: 70179645-70179645 on Build GRCh38
rs777665519 Chr.2: 70179655-70179655 on Build GRCh38
rs755152746 Chr.2: 70179651-70179651 on Build GRCh38

View Full Product Details