Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66493296-66493521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEAN1
Entrez Gene ID
146227
Gene Name
brain expressed, associated with NEDD4, 1
Gene Aliases
BEAN, SCA31
UniGene
Hs.740218
Gene Chromosome Location
Chr.16: 66427297-66495288 on Build GRCh38
Transcripts
NM_001197224.3 NM_001197225.3

SNP Detail

SNP ID Chromosome Location
rs946911210 Chr.16: 66493337-66493337 on Build GRCh38
rs117499742 Chr.16: 66493333-66493333 on Build GRCh38
rs762955232 Chr.16: 66493332-66493332 on Build GRCh38
rs199506725 Chr.16: 66493454-66493454 on Build GRCh38
rs534486281 Chr.16: 66493447-66493447 on Build GRCh38
rs1051956841 Chr.16: 66493428-66493428 on Build GRCh38
rs572041597 Chr.16: 66493421-66493421 on Build GRCh38
rs1017304429 Chr.16: 66493495-66493495 on Build GRCh38
rs762129438 Chr.16: 66493355-66493355 on Build GRCh38
rs753648974 Chr.16: 66493455-66493455 on Build GRCh38
rs1051124243 Chr.16: 66493457-66493457 on Build GRCh38
rs370820703 Chr.16: 66493350-66493350 on Build GRCh38
rs143061241 Chr.16: 66493353-66493353 on Build GRCh38
rs139137875 Chr.16: 66493328-66493328 on Build GRCh38
rs890085665 Chr.16: 66493443-66493443 on Build GRCh38
rs763515994 Chr.16: 66493374-66493374 on Build GRCh38
rs767675949 Chr.16: 66493388-66493388 on Build GRCh38
rs564938855 Chr.16: 66493443-66493444 on Build GRCh38
rs190111095 Chr.16: 66493489-66493489 on Build GRCh38

View Full Product Details