Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116401016-116401289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A1
Entrez Gene ID
476
Gene Name
ATPase Na+/K+ transporting subunit alpha 1
Gene Aliases
-
UniGene
Hs.371889
Gene Chromosome Location
Chr.1: 116373173-116404774 on Build GRCh38
Transcripts
NM_000701.7 NM_001160233.1 NM_001160234.1 XM_017001360.1 XM_017001361.1 NR_027645.1 NR_027646.1

Gene Symbol
ATP1A1-AS1
Entrez Gene ID
84852
Gene Name
ATP1A1 antisense RNA 1
Gene Aliases
ATP1A1OS, C1orf203
UniGene
Hs.664752
Gene Chromosome Location
Chr.1: 116392865-116418622 on Build GRCh38
Transcripts
NM_000701.7 NM_001160233.1 NM_001160234.1 XM_017001360.1 XM_017001361.1 NR_027645.1 NR_027646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1333026 c.2740G>A p.V914M Chr.1: 116401151-116401151 on Build GRCh38
5498744 c.2796C>T p.A932A Chr.1: 116401207-116401207 on Build GRCh38
172905 c.2769C>T p.F923F Chr.1: 116401180-116401180 on Build GRCh38
3710026 c.2719-1G>C p.? Chr.1: 116401129-116401129 on Build GRCh38
6331674 c.2828C>T p.S943L Chr.1: 116401239-116401239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757409362 Chr.1: 116401044-116401044 on Build GRCh38
rs762463400 Chr.1: 116401122-116401122 on Build GRCh38
rs763654501 Chr.1: 116401132-116401132 on Build GRCh38
rs757162609 Chr.1: 116401168-116401168 on Build GRCh38
rs745657240 Chr.1: 116401180-116401180 on Build GRCh38
rs759943783 Chr.1: 116401243-116401243 on Build GRCh38
rs138167001 Chr.1: 116401240-116401240 on Build GRCh38
rs111965968 Chr.1: 116401262-116401262 on Build GRCh38
rs905279171 Chr.1: 116401108-116401108 on Build GRCh38
rs765387548 Chr.1: 116401155-116401155 on Build GRCh38
rs140795898 Chr.1: 116401159-116401159 on Build GRCh38
rs755301003 Chr.1: 116401153-116401153 on Build GRCh38
rs765834451 Chr.1: 116401038-116401039 on Build GRCh38
rs749010651 Chr.1: 116401195-116401195 on Build GRCh38
rs769364704 Chr.1: 116401107-116401107 on Build GRCh38
rs192890533 Chr.1: 116401083-116401083 on Build GRCh38
rs747010288 Chr.1: 116401089-116401089 on Build GRCh38
rs11540950 Chr.1: 116401139-116401139 on Build GRCh38
rs373693545 Chr.1: 116401138-116401138 on Build GRCh38
rs753079721 Chr.1: 116401263-116401263 on Build GRCh38
rs755175168 Chr.1: 116401035-116401035 on Build GRCh38
rs775056444 Chr.1: 116401109-116401109 on Build GRCh38
rs114525271 Chr.1: 116401150-116401150 on Build GRCh38
rs777986651 Chr.1: 116401085-116401085 on Build GRCh38
rs937122853 Chr.1: 116401135-116401135 on Build GRCh38
rs773902417 Chr.1: 116401211-116401211 on Build GRCh38
rs761343155 Chr.1: 116401214-116401214 on Build GRCh38
rs747061651 Chr.1: 116401160-116401160 on Build GRCh38
rs753022560 Chr.1: 116401158-116401158 on Build GRCh38
rs781145047 Chr.1: 116401174-116401174 on Build GRCh38
rs748238972 Chr.1: 116401079-116401079 on Build GRCh38
rs771412029 Chr.1: 116401096-116401096 on Build GRCh38
rs112588853 Chr.1: 116401128-116401128 on Build GRCh38
rs570205727 Chr.1: 116401207-116401207 on Build GRCh38
rs111567351 Chr.1: 116401105-116401105 on Build GRCh38
rs765762308 Chr.1: 116401254-116401254 on Build GRCh38
rs763313496 Chr.1: 116401265-116401265 on Build GRCh38
rs769574702 Chr.1: 116401189-116401189 on Build GRCh38
rs904188232 Chr.1: 116401255-116401255 on Build GRCh38
rs770869370 Chr.1: 116401090-116401090 on Build GRCh38
rs775252649 Chr.1: 116401190-116401190 on Build GRCh38
rs941392154 Chr.1: 116401067-116401067 on Build GRCh38

View Full Product Details