Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120408292-120408533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5RAP2
Entrez Gene ID
55755
Gene Name
CDK5 regulatory subunit associated protein 2
Gene Aliases
C48, Cep215, MCPH3
UniGene
Hs.269560
Gene Chromosome Location
Chr.9: 120388869-120580170 on Build GRCh38
Transcripts
NM_001011649.2 NM_001272039.1 NM_018249.5 NR_073554.1 NR_073555.1 NR_073556.1 NR_073557.1 NR_073558.1 XM_006717182.1 XM_006717185.1 XM_011518860.1 XM_017014921.1 XM_017014922.1 XM_017014923.1 XM_017014924.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6182483 c.4683G>A p.L1561L Chr.9: 120408390-120408390 on Build GRCh38
1314406 c.4712A>T p.H1571L Chr.9: 120408361-120408361 on Build GRCh38
6625559 c.4643C>T p.S1548L Chr.9: 120408430-120408430 on Build GRCh38
5003166 c.4618G>C p.V1540L Chr.9: 120408455-120408455 on Build GRCh38
6625564 c.4672C>T p.R1558* Chr.9: 120408401-120408401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017376531 Chr.9: 120408393-120408393 on Build GRCh38
rs764765925 Chr.9: 120408469-120408469 on Build GRCh38
rs756634521 Chr.9: 120408483-120408483 on Build GRCh38
rs767068770 Chr.9: 120408478-120408478 on Build GRCh38
rs778986149 Chr.9: 120408500-120408500 on Build GRCh38
rs776070446 Chr.9: 120408458-120408458 on Build GRCh38
rs755285807 Chr.9: 120408476-120408476 on Build GRCh38
rs752004121 Chr.9: 120408474-120408474 on Build GRCh38
rs1033138156 Chr.9: 120408414-120408414 on Build GRCh38
rs781529498 Chr.9: 120408390-120408390 on Build GRCh38
rs778026517 Chr.9: 120408492-120408492 on Build GRCh38
rs763605383 Chr.9: 120408457-120408457 on Build GRCh38
rs140993056 Chr.9: 120408434-120408434 on Build GRCh38
rs749343479 Chr.9: 120408493-120408493 on Build GRCh38
rs527666804 Chr.9: 120408363-120408363 on Build GRCh38
rs748298248 Chr.9: 120408392-120408392 on Build GRCh38
rs979590267 Chr.9: 120408382-120408382 on Build GRCh38
rs761557621 Chr.9: 120408468-120408468 on Build GRCh38
rs193196156 Chr.9: 120408479-120408479 on Build GRCh38
rs762201727 Chr.9: 120408411-120408412 on Build GRCh38
rs1052699135 Chr.9: 120408432-120408432 on Build GRCh38
rs775025723 Chr.9: 120408315-120408315 on Build GRCh38
rs766788348 Chr.9: 120408364-120408364 on Build GRCh38
rs775398273 Chr.9: 120408450-120408450 on Build GRCh38
rs373278668 Chr.9: 120408401-120408401 on Build GRCh38
rs891410760 Chr.9: 120408436-120408436 on Build GRCh38
rs763799606 Chr.9: 120408329-120408329 on Build GRCh38
rs753263727 Chr.9: 120408481-120408481 on Build GRCh38
rs757544827 Chr.9: 120408499-120408499 on Build GRCh38
rs141563261 Chr.9: 120408447-120408447 on Build GRCh38
rs35909061 Chr.9: 120408408-120408408 on Build GRCh38
rs4837768 Chr.9: 120408455-120408455 on Build GRCh38
rs753492465 Chr.9: 120408344-120408344 on Build GRCh38
rs771014048 Chr.9: 120408426-120408426 on Build GRCh38
rs772102203 Chr.9: 120408449-120408449 on Build GRCh38
rs755549517 Chr.9: 120408385-120408385 on Build GRCh38
rs899576554 Chr.9: 120408467-120408467 on Build GRCh38
rs948169747 Chr.9: 120408380-120408380 on Build GRCh38
rs373837579 Chr.9: 120408501-120408501 on Build GRCh38
rs377338416 Chr.9: 120408400-120408400 on Build GRCh38
rs761191015 Chr.9: 120408477-120408477 on Build GRCh38
rs751830557 Chr.9: 120408379-120408379 on Build GRCh38
rs760162988 Chr.9: 120408322-120408322 on Build GRCh38
rs560388981 Chr.9: 120408352-120408352 on Build GRCh38
rs1048188142 Chr.9: 120408486-120408486 on Build GRCh38

View Full Product Details