Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 107051014-107051285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf203
Entrez Gene ID
51250
Gene Name
chromosome 6 open reading frame 203
Gene Aliases
HSPC230, PRED31
UniGene
Hs.486084
Gene Chromosome Location
Chr.6: 107028172-107051586 on Build GRCh38
Transcripts
NM_001142468.2 NM_001142470.2 NM_016487.4 XM_011535876.2 XM_011535877.2 XM_017010919.1 XM_017010920.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3429826 c.665G>T p.R222I Chr.6: 107051178-107051178 on Build GRCh38
5419009 c.637_641delGAAGA p.E214fs*2 Chr.6: 107051150-107051154 on Build GRCh38
7209361 c.642_646delGAAGA p.E214fs*2 Chr.6: 107051155-107051159 on Build GRCh38
1246924 c.675_676insCGG p.R227_W228insR Chr.6: 107051188-107051189 on Build GRCh38
239087 c.630_631insA p.V211fs*3 Chr.6: 107051143-107051144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031818945 Chr.6: 107051103-107051103 on Build GRCh38
rs919766391 Chr.6: 107051215-107051215 on Build GRCh38
rs782131098 Chr.6: 107051209-107051209 on Build GRCh38
rs781867556 Chr.6: 107051120-107051120 on Build GRCh38
rs915857922 Chr.6: 107051161-107051161 on Build GRCh38
rs375132159 Chr.6: 107051074-107051074 on Build GRCh38
rs782434105 Chr.6: 107051195-107051195 on Build GRCh38
rs782194070 Chr.6: 107051044-107051044 on Build GRCh38
rs782666487 Chr.6: 107051126-107051126 on Build GRCh38
rs562109891 Chr.6: 107051088-107051088 on Build GRCh38
rs530002315 Chr.6: 107051192-107051192 on Build GRCh38
rs782513064 Chr.6: 107051124-107051124 on Build GRCh38
rs782704372 Chr.6: 107051203-107051203 on Build GRCh38
rs782308531 Chr.6: 107051159-107051160 on Build GRCh38
rs782162548 Chr.6: 107051081-107051081 on Build GRCh38
rs782071029 Chr.6: 107051201-107051201 on Build GRCh38
rs781945681 Chr.6: 107051206-107051206 on Build GRCh38
rs200460291 Chr.6: 107051133-107051133 on Build GRCh38
rs782278945 Chr.6: 107051177-107051178 on Build GRCh38
rs200241235 Chr.6: 107051188-107051188 on Build GRCh38
rs146122461 Chr.6: 107051053-107051053 on Build GRCh38
rs185499979 Chr.6: 107051178-107051178 on Build GRCh38
rs782722011 Chr.6: 107051232-107051232 on Build GRCh38
rs981879224 Chr.6: 107051182-107051182 on Build GRCh38
rs782728814 Chr.6: 107051114-107051114 on Build GRCh38
rs782064050 Chr.6: 107051038-107051039 on Build GRCh38
rs781986540 Chr.6: 107051080-107051080 on Build GRCh38
rs199681142 Chr.6: 107051214-107051214 on Build GRCh38
rs199936497 Chr.6: 107051043-107051043 on Build GRCh38
rs782349808 Chr.6: 107051190-107051190 on Build GRCh38
rs202118375 Chr.6: 107051073-107051073 on Build GRCh38
rs200587674 Chr.6: 107051198-107051198 on Build GRCh38
rs781810219 Chr.6: 107051091-107051091 on Build GRCh38
rs782483224 Chr.6: 107051095-107051095 on Build GRCh38
rs531713964 Chr.6: 107051143-107051143 on Build GRCh38
rs562107376 Chr.6: 107051186-107051186 on Build GRCh38
rs782521747 Chr.6: 107051218-107051218 on Build GRCh38
rs781973254 Chr.6: 107051060-107051060 on Build GRCh38
rs939939252 Chr.6: 107051202-107051202 on Build GRCh38
rs532303825 Chr.6: 107051246-107051246 on Build GRCh38
rs369906226 Chr.6: 107051244-107051244 on Build GRCh38
rs140099248 Chr.6: 107051193-107051193 on Build GRCh38
rs782752094 Chr.6: 107051212-107051212 on Build GRCh38
rs782494174 Chr.6: 107051245-107051245 on Build GRCh38
rs201067031 Chr.6: 107051077-107051077 on Build GRCh38
rs367978912 Chr.6: 107051127-107051127 on Build GRCh38

View Full Product Details