Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45620684-45620911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC17
Entrez Gene ID
149483
Gene Name
coiled-coil domain containing 17
Gene Aliases
-
UniGene
Hs.18912
Gene Chromosome Location
Chr.1: 45620044-45624787 on Build GRCh38
Transcripts
NM_001114938.2 NM_001190182.1 XM_011540812.1 XM_011540814.1 XM_017000450.1 XM_017000451.1 XM_017000452.1 XM_017000453.1 XM_017000454.1 XM_017000455.1 XM_017000456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3360928 c.1560G>C p.Q520H Chr.1: 45620777-45620777 on Build GRCh38
5832608 c.1601_1602insT p.Q534fs*25 Chr.1: 45620735-45620736 on Build GRCh38
3400838 c.1503G>A p.Q501Q Chr.1: 45620903-45620903 on Build GRCh38
6417229 c.1607C>A p.P536Q Chr.1: 45620730-45620730 on Build GRCh38
4165211 c.1550C>T p.A517V Chr.1: 45620787-45620787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752566827 Chr.1: 45620800-45620800 on Build GRCh38
rs749570663 Chr.1: 45620839-45620840 on Build GRCh38
rs1048067280 Chr.1: 45620848-45620848 on Build GRCh38
rs781635850 Chr.1: 45620852-45620852 on Build GRCh38
rs756245036 Chr.1: 45620794-45620795 on Build GRCh38
rs755794667 Chr.1: 45620802-45620802 on Build GRCh38
rs908853305 Chr.1: 45620860-45620860 on Build GRCh38
rs984407882 Chr.1: 45620845-45620845 on Build GRCh38
rs769997849 Chr.1: 45620863-45620863 on Build GRCh38
rs369435540 Chr.1: 45620808-45620808 on Build GRCh38
rs747514252 Chr.1: 45620745-45620745 on Build GRCh38
rs771183008 Chr.1: 45620879-45620879 on Build GRCh38
rs759644586 Chr.1: 45620884-45620884 on Build GRCh38
rs2297654 Chr.1: 45620764-45620764 on Build GRCh38
rs764795876 Chr.1: 45620809-45620809 on Build GRCh38
rs770086992 Chr.1: 45620755-45620755 on Build GRCh38
rs374255983 Chr.1: 45620890-45620890 on Build GRCh38
rs781102815 Chr.1: 45620714-45620714 on Build GRCh38
rs112139251 Chr.1: 45620778-45620778 on Build GRCh38
rs755808620 Chr.1: 45620724-45620724 on Build GRCh38
rs752667473 Chr.1: 45620735-45620754 on Build GRCh38
rs756861130 Chr.1: 45620812-45620812 on Build GRCh38
rs752446141 Chr.1: 45620722-45620722 on Build GRCh38
rs769053033 Chr.1: 45620749-45620749 on Build GRCh38
rs779485254 Chr.1: 45620867-45620867 on Build GRCh38
rs181566146 Chr.1: 45620883-45620883 on Build GRCh38
rs566975492 Chr.1: 45620751-45620751 on Build GRCh38
rs778106401 Chr.1: 45620815-45620816 on Build GRCh38
rs763149404 Chr.1: 45620767-45620767 on Build GRCh38
rs773548306 Chr.1: 45620869-45620869 on Build GRCh38
rs778366726 Chr.1: 45620815-45620815 on Build GRCh38
rs754886430 Chr.1: 45620704-45620704 on Build GRCh38
rs762639663 Chr.1: 45620889-45620889 on Build GRCh38
rs202073754 Chr.1: 45620797-45620797 on Build GRCh38
rs958387687 Chr.1: 45620835-45620835 on Build GRCh38
rs796836865 Chr.1: 45620717-45620717 on Build GRCh38
rs755503077 Chr.1: 45620839-45620839 on Build GRCh38
rs772403552 Chr.1: 45620891-45620891 on Build GRCh38
rs771407862 Chr.1: 45620860-45620861 on Build GRCh38
rs747567705 Chr.1: 45620834-45620834 on Build GRCh38
rs746246563 Chr.1: 45620871-45620872 on Build GRCh38
rs748414023 Chr.1: 45620752-45620752 on Build GRCh38
rs143313832 Chr.1: 45620735-45620735 on Build GRCh38
rs530067462 Chr.1: 45620732-45620732 on Build GRCh38
rs748552146 Chr.1: 45620854-45620854 on Build GRCh38
rs775301308 Chr.1: 45620870-45620870 on Build GRCh38
rs766364006 Chr.1: 45620772-45620772 on Build GRCh38
rs774469215 Chr.1: 45620781-45620781 on Build GRCh38

View Full Product Details