Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10929476-10929737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf52
Entrez Gene ID
90580
Gene Name
chromosome 19 open reading frame 52
Gene Aliases
-
UniGene
Hs.164026
Gene Chromosome Location
Chr.19: 10928748-10930241 on Build GRCh38
Transcripts
NM_138358.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6319111 c.702G>T p.E234D Chr.19: 10929621-10929621 on Build GRCh38
1303965 c.723G>A p.K241K Chr.19: 10929642-10929642 on Build GRCh38
4154036 c.574C>T p.L192L Chr.19: 10929493-10929493 on Build GRCh38
239033 c.668C>G p.T223S Chr.19: 10929587-10929587 on Build GRCh38
4557173 c.717G>A p.K239K Chr.19: 10929636-10929636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923414662 Chr.19: 10929609-10929609 on Build GRCh38
rs144191264 Chr.19: 10929593-10929593 on Build GRCh38
rs780304065 Chr.19: 10929619-10929619 on Build GRCh38
rs762059274 Chr.19: 10929502-10929502 on Build GRCh38
rs775422701 Chr.19: 10929658-10929658 on Build GRCh38
rs770004591 Chr.19: 10929636-10929636 on Build GRCh38
rs1005301767 Chr.19: 10929504-10929504 on Build GRCh38
rs117203028 Chr.19: 10929587-10929587 on Build GRCh38
rs971196198 Chr.19: 10929699-10929699 on Build GRCh38
rs1027960252 Chr.19: 10929495-10929495 on Build GRCh38
rs761677271 Chr.19: 10929638-10929640 on Build GRCh38
rs753435701 Chr.19: 10929607-10929607 on Build GRCh38
rs750039216 Chr.19: 10929695-10929695 on Build GRCh38
rs751506909 Chr.19: 10929510-10929510 on Build GRCh38
rs747449095 Chr.19: 10929622-10929622 on Build GRCh38
rs750638662 Chr.19: 10929547-10929547 on Build GRCh38
rs371423456 Chr.19: 10929571-10929571 on Build GRCh38
rs374504443 Chr.19: 10929578-10929578 on Build GRCh38
rs764977165 Chr.19: 10929694-10929694 on Build GRCh38
rs752257479 Chr.19: 10929557-10929557 on Build GRCh38
rs750193226 Chr.19: 10929508-10929508 on Build GRCh38
rs35087899 Chr.19: 10929696-10929696 on Build GRCh38
rs775920498 Chr.19: 10929539-10929539 on Build GRCh38
rs555565275 Chr.19: 10929516-10929516 on Build GRCh38
rs754907453 Chr.19: 10929553-10929553 on Build GRCh38
rs754501820 Chr.19: 10929514-10929514 on Build GRCh38
rs1055837080 Chr.19: 10929711-10929732 on Build GRCh38
rs367796118 Chr.19: 10929633-10929633 on Build GRCh38
rs967601462 Chr.19: 10929634-10929634 on Build GRCh38
rs753259354 Chr.19: 10929707-10929707 on Build GRCh38
rs763538420 Chr.19: 10929605-10929605 on Build GRCh38
rs747500580 Chr.19: 10929573-10929573 on Build GRCh38
rs781780438 Chr.19: 10929632-10929632 on Build GRCh38
rs749496771 Chr.19: 10929639-10929639 on Build GRCh38
rs755591004 Chr.19: 10929558-10929558 on Build GRCh38
rs994030404 Chr.19: 10929684-10929684 on Build GRCh38
rs765153717 Chr.19: 10929505-10929505 on Build GRCh38
rs751514423 Chr.19: 10929712-10929713 on Build GRCh38
rs764775842 Chr.19: 10929554-10929560 on Build GRCh38
rs746259617 Chr.19: 10929536-10929536 on Build GRCh38
rs767040521 Chr.19: 10929555-10929555 on Build GRCh38
rs750912802 Chr.19: 10929509-10929509 on Build GRCh38
rs755359545 Chr.19: 10929630-10929630 on Build GRCh38
rs202159235 Chr.19: 10929501-10929501 on Build GRCh38
rs761237926 Chr.19: 10929526-10929526 on Build GRCh38
rs759396827 Chr.19: 10929647-10929647 on Build GRCh38
rs375480345 Chr.19: 10929543-10929543 on Build GRCh38
rs761183840 Chr.19: 10929540-10929540 on Build GRCh38
rs150842814 Chr.19: 10929580-10929580 on Build GRCh38
rs141741045 Chr.19: 10929549-10929549 on Build GRCh38
rs760541406 Chr.19: 10929669-10929669 on Build GRCh38
rs761202594 Chr.19: 10929691-10929691 on Build GRCh38
rs760807134 Chr.19: 10929519-10929519 on Build GRCh38
rs747832687 Chr.19: 10929520-10929520 on Build GRCh38
rs773361746 Chr.19: 10929579-10929579 on Build GRCh38
rs777255213 Chr.19: 10929575-10929575 on Build GRCh38
rs200200202 Chr.19: 10929537-10929537 on Build GRCh38
rs763541690 Chr.19: 10929680-10929680 on Build GRCh38
rs775396605 Chr.19: 10929608-10929608 on Build GRCh38
rs377637993 Chr.19: 10929550-10929550 on Build GRCh38
rs544129891 Chr.19: 10929559-10929559 on Build GRCh38
rs773305467 Chr.19: 10929637-10929637 on Build GRCh38
rs1038159593 Chr.19: 10929655-10929655 on Build GRCh38
rs777386752 Chr.19: 10929521-10929521 on Build GRCh38
rs748958833 Chr.19: 10929570-10929570 on Build GRCh38
rs767486530 Chr.19: 10929588-10929588 on Build GRCh38
rs774410164 Chr.19: 10929643-10929643 on Build GRCh38
rs756891932 Chr.19: 10929616-10929616 on Build GRCh38
rs765601577 Chr.19: 10929546-10929546 on Build GRCh38
rs769236430 Chr.19: 10929574-10929574 on Build GRCh38
rs146558038 Chr.19: 10929687-10929687 on Build GRCh38
rs751458356 Chr.19: 10929552-10929552 on Build GRCh38
rs139319173 Chr.19: 10929592-10929592 on Build GRCh38
rs549183212 Chr.19: 10929716-10929716 on Build GRCh38
rs544987672 Chr.19: 10929642-10929642 on Build GRCh38
rs759319722 Chr.19: 10929539-10929571 on Build GRCh38
rs774150261 Chr.19: 10929581-10929581 on Build GRCh38
rs762689957 Chr.19: 10929635-10929637 on Build GRCh38
rs371039420 Chr.19: 10929513-10929513 on Build GRCh38

View Full Product Details