Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57246045-57246308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL2BP
Entrez Gene ID
23568
Gene Name
ADP ribosylation factor like GTPase 2 binding protein
Gene Aliases
BART, BART1, RP66
UniGene
Hs.632873
Gene Chromosome Location
Chr.16: 57245126-57253635 on Build GRCh38
Transcripts
NM_012106.3 XM_011522977.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4495747 c.45C>T p.S15S Chr.16: 57246086-57246086 on Build GRCh38
4982209 c.64G>C p.D22H Chr.16: 57246105-57246105 on Build GRCh38
3679928 c.39-9G>T p.? Chr.16: 57246071-57246071 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758680473 Chr.16: 57246145-57246145 on Build GRCh38
rs773407657 Chr.16: 57246093-57246093 on Build GRCh38
rs772330581 Chr.16: 57246203-57246203 on Build GRCh38
rs111658181 Chr.16: 57246098-57246098 on Build GRCh38
rs781043409 Chr.16: 57246157-57246157 on Build GRCh38
rs113676384 Chr.16: 57246164-57246164 on Build GRCh38
rs141614381 Chr.16: 57246138-57246138 on Build GRCh38
rs754369850 Chr.16: 57246130-57246130 on Build GRCh38
rs771739790 Chr.16: 57246193-57246193 on Build GRCh38
rs549381507 Chr.16: 57246202-57246202 on Build GRCh38
rs750070601 Chr.16: 57246159-57246159 on Build GRCh38
rs199762075 Chr.16: 57246135-57246135 on Build GRCh38
rs755458529 Chr.16: 57246131-57246131 on Build GRCh38
rs757449294 Chr.16: 57246137-57246138 on Build GRCh38
rs369932682 Chr.16: 57246182-57246182 on Build GRCh38
rs530042012 Chr.16: 57246172-57246172 on Build GRCh38
rs370696038 Chr.16: 57246188-57246188 on Build GRCh38
rs749196911 Chr.16: 57246176-57246176 on Build GRCh38
rs779663179 Chr.16: 57246175-57246175 on Build GRCh38
rs148438260 Chr.16: 57246099-57246099 on Build GRCh38
rs766410828 Chr.16: 57246106-57246106 on Build GRCh38
rs957928948 Chr.16: 57246197-57246197 on Build GRCh38
rs748031120 Chr.16: 57246192-57246192 on Build GRCh38

View Full Product Details