Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33912158-33912401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5O
Entrez Gene ID
539
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit
Gene Aliases
ATPO, HMC08D05, OSCP
UniGene
Hs.409140
Gene Chromosome Location
Chr.21: 33903453-33915854 on Build GRCh38
Transcripts
NM_001697.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
73778 c.190A>G p.R64G Chr.21: 33912297-33912297 on Build GRCh38
6467918 c.88-1_95delGCCTCCTGT p.? Chr.21: 33912392-33912400 on Build GRCh38
6458915 c.97_98insAAGAGACAA p.Q33_V34insETK Chr.21: 33912389-33912390 on Build GRCh38
7263530 c.130G>T p.A44S Chr.21: 33912357-33912357 on Build GRCh38
5392856 c.125C>T p.A42V Chr.21: 33912362-33912362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368448400 Chr.21: 33912250-33912250 on Build GRCh38
rs1054339303 Chr.21: 33912371-33912371 on Build GRCh38
rs776875621 Chr.21: 33912319-33912319 on Build GRCh38
rs949072683 Chr.21: 33912254-33912254 on Build GRCh38
rs941809143 Chr.21: 33912185-33912185 on Build GRCh38
rs772564305 Chr.21: 33912288-33912288 on Build GRCh38
rs1042613037 Chr.21: 33912264-33912264 on Build GRCh38
rs80010393 Chr.21: 33912261-33912261 on Build GRCh38
rs114179065 Chr.21: 33912201-33912201 on Build GRCh38
rs78234759 Chr.21: 33912379-33912379 on Build GRCh38
rs2834299 Chr.21: 33912198-33912198 on Build GRCh38
rs775920539 Chr.21: 33912309-33912309 on Build GRCh38
rs764371658 Chr.21: 33912314-33912314 on Build GRCh38
rs763837966 Chr.21: 33912316-33912316 on Build GRCh38
rs553480576 Chr.21: 33912221-33912221 on Build GRCh38
rs990760109 Chr.21: 33912226-33912226 on Build GRCh38
rs762153291 Chr.21: 33912363-33912363 on Build GRCh38
rs1000967266 Chr.21: 33912368-33912368 on Build GRCh38
rs557551084 Chr.21: 33912188-33912188 on Build GRCh38
rs974156334 Chr.21: 33912186-33912186 on Build GRCh38
rs762497837 Chr.21: 33912275-33912275 on Build GRCh38
rs765607316 Chr.21: 33912364-33912364 on Build GRCh38
rs937886291 Chr.21: 33912209-33912209 on Build GRCh38
rs79871106 Chr.21: 33912338-33912338 on Build GRCh38
rs199872694 Chr.21: 33912258-33912258 on Build GRCh38

View Full Product Details