Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 56563961-56564210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC66
Entrez Gene ID
285331
Gene Name
coiled-coil domain containing 66
Gene Aliases
-
UniGene
Hs.476399
Gene Chromosome Location
Chr.3: 56557125-56621837 on Build GRCh38
Transcripts
NM_001012506.4 NM_001141947.1 NR_024460.1 XM_005265081.3 XM_005265082.3 XM_005265083.3 XM_005265084.3 XM_011533614.2 XM_011533615.2 XM_011533616.2 XM_011533617.2 XM_011533619.2 XM_017006229.1 XM_017006230.1 XM_017006231.1 XM_017006232.1 XM_017006233.1 XM_017006234.1 XM_017006235.1 XM_017006236.1 XM_017006237.1 XM_017006238.1 XM_017006240.1 XM_017006241.1 XM_017006242.1 XM_017006243.1 XM_017006244.1 XM_017006245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1186327 c.57C>A p.H19Q Chr.3: 56563989-56563989 on Build GRCh38
6622354 c.520A>G p.T174A Chr.3: 56564101-56564101 on Build GRCh38
4337507 c.409delA p.K138fs*8 Chr.3: 56563990-56563990 on Build GRCh38
3767500 c.168G>C p.L56L Chr.3: 56564100-56564100 on Build GRCh38
1566666 c.58delA p.K21fs*8 Chr.3: 56563990-56563990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747483025 Chr.3: 56564170-56564170 on Build GRCh38
rs376747220 Chr.3: 56564004-56564004 on Build GRCh38
rs745683890 Chr.3: 56564033-56564033 on Build GRCh38
rs143579084 Chr.3: 56564108-56564108 on Build GRCh38
rs371171885 Chr.3: 56564144-56564144 on Build GRCh38
rs144464888 Chr.3: 56564090-56564090 on Build GRCh38
rs775010206 Chr.3: 56564065-56564065 on Build GRCh38
rs938966971 Chr.3: 56564166-56564166 on Build GRCh38
rs534153650 Chr.3: 56564077-56564077 on Build GRCh38
rs868169218 Chr.3: 56564097-56564097 on Build GRCh38
rs768728396 Chr.3: 56563994-56563994 on Build GRCh38
rs748201227 Chr.3: 56564047-56564047 on Build GRCh38
rs767995749 Chr.3: 56564006-56564006 on Build GRCh38
rs373624834 Chr.3: 56563999-56563999 on Build GRCh38
rs762724461 Chr.3: 56564069-56564069 on Build GRCh38
rs781246405 Chr.3: 56564031-56564031 on Build GRCh38
rs772605017 Chr.3: 56564053-56564053 on Build GRCh38
rs1046614453 Chr.3: 56564093-56564093 on Build GRCh38
rs9848723 Chr.3: 56564148-56564148 on Build GRCh38
rs780157815 Chr.3: 56564040-56564040 on Build GRCh38
rs891868770 Chr.3: 56563989-56563989 on Build GRCh38
rs751101219 Chr.3: 56564027-56564027 on Build GRCh38
rs773613935 Chr.3: 56564012-56564012 on Build GRCh38
rs141268275 Chr.3: 56564106-56564106 on Build GRCh38
rs775743963 Chr.3: 56563990-56563990 on Build GRCh38
rs551819797 Chr.3: 56564014-56564014 on Build GRCh38
rs779962162 Chr.3: 56564118-56564118 on Build GRCh38
rs992938109 Chr.3: 56564167-56564167 on Build GRCh38
rs371128265 Chr.3: 56564038-56564038 on Build GRCh38
rs773726330 Chr.3: 56564056-56564056 on Build GRCh38
rs571674487 Chr.3: 56564062-56564062 on Build GRCh38
rs773604990 Chr.3: 56564149-56564149 on Build GRCh38
rs763816920 Chr.3: 56564070-56564070 on Build GRCh38
rs749019281 Chr.3: 56564126-56564126 on Build GRCh38
rs267599912 Chr.3: 56564122-56564122 on Build GRCh38
rs371159686 Chr.3: 56564063-56564063 on Build GRCh38
rs755870203 Chr.3: 56564109-56564109 on Build GRCh38
rs774469386 Chr.3: 56564043-56564043 on Build GRCh38
rs763763362 Chr.3: 56564023-56564023 on Build GRCh38
rs142753279 Chr.3: 56563991-56563991 on Build GRCh38
rs768856982 Chr.3: 56564041-56564041 on Build GRCh38
rs757859135 Chr.3: 56564022-56564022 on Build GRCh38
rs1019486021 Chr.3: 56564084-56564084 on Build GRCh38
rs75201116 Chr.3: 56564013-56564013 on Build GRCh38
rs752344741 Chr.3: 56564016-56564016 on Build GRCh38
rs774839605 Chr.3: 56563996-56563996 on Build GRCh38
rs377469116 Chr.3: 56564139-56564139 on Build GRCh38
rs754989007 Chr.3: 56564158-56564159 on Build GRCh38

View Full Product Details