Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 105115625-105115898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BVES
Entrez Gene ID
11149
Gene Name
blood vessel epicardial substance
Gene Aliases
HBVES, LGMD2X, POP1, POPDC1
UniGene
Hs.221660
Gene Chromosome Location
Chr.6: 105096822-105137174 on Build GRCh38
Transcripts
NM_001199563.1 NM_007073.4 NM_147147.3 XM_011535398.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1739289 c.828G>C p.K276N Chr.6: 105115816-105115816 on Build GRCh38
3857219 c.851G>A p.C284Y Chr.6: 105115793-105115793 on Build GRCh38
4510753 c.847C>T p.L283F Chr.6: 105115797-105115797 on Build GRCh38
5457214 c.833A>G p.E278G Chr.6: 105115811-105115811 on Build GRCh38
377971 c.944G>A p.S315N Chr.6: 105115700-105115700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564443311 Chr.6: 105115834-105115834 on Build GRCh38
rs752058250 Chr.6: 105115800-105115800 on Build GRCh38
rs547362212 Chr.6: 105115707-105115707 on Build GRCh38
rs185464255 Chr.6: 105115735-105115735 on Build GRCh38
rs984356256 Chr.6: 105115778-105115778 on Build GRCh38
rs755973295 Chr.6: 105115724-105115724 on Build GRCh38
rs1004017848 Chr.6: 105115762-105115762 on Build GRCh38
rs964037211 Chr.6: 105115803-105115803 on Build GRCh38
rs747087256 Chr.6: 105115743-105115743 on Build GRCh38
rs921674998 Chr.6: 105115700-105115700 on Build GRCh38
rs753635426 Chr.6: 105115847-105115847 on Build GRCh38
rs765528545 Chr.6: 105115759-105115759 on Build GRCh38
rs745871502 Chr.6: 105115870-105115870 on Build GRCh38
rs940897167 Chr.6: 105115731-105115731 on Build GRCh38
rs370149373 Chr.6: 105115809-105115809 on Build GRCh38
rs773168306 Chr.6: 105115739-105115739 on Build GRCh38
rs774193505 Chr.6: 105115742-105115742 on Build GRCh38
rs41285458 Chr.6: 105115802-105115802 on Build GRCh38
rs760559899 Chr.6: 105115837-105115838 on Build GRCh38
rs139953651 Chr.6: 105115750-105115750 on Build GRCh38
rs922662605 Chr.6: 105115839-105115839 on Build GRCh38
rs775658248 Chr.6: 105115658-105115658 on Build GRCh38
rs773650318 Chr.6: 105115777-105115777 on Build GRCh38
rs762150631 Chr.6: 105115673-105115673 on Build GRCh38
rs374455938 Chr.6: 105115789-105115789 on Build GRCh38
rs371189369 Chr.6: 105115713-105115713 on Build GRCh38
rs767206976 Chr.6: 105115808-105115808 on Build GRCh38
rs192880685 Chr.6: 105115676-105115676 on Build GRCh38
rs368905975 Chr.6: 105115655-105115655 on Build GRCh38
rs930623402 Chr.6: 105115738-105115738 on Build GRCh38
rs776828424 Chr.6: 105115745-105115745 on Build GRCh38
rs760766482 Chr.6: 105115659-105115659 on Build GRCh38
rs374353418 Chr.6: 105115859-105115859 on Build GRCh38
rs768780960 Chr.6: 105115744-105115744 on Build GRCh38
rs772041983 Chr.6: 105115663-105115664 on Build GRCh38
rs541771381 Chr.6: 105115712-105115712 on Build GRCh38
rs934066932 Chr.6: 105115843-105115843 on Build GRCh38
rs147469762 Chr.6: 105115734-105115734 on Build GRCh38
rs764267987 Chr.6: 105115661-105115661 on Build GRCh38
rs776886182 Chr.6: 105115672-105115672 on Build GRCh38
rs865878316 Chr.6: 105115765-105115765 on Build GRCh38
rs1035150303 Chr.6: 105115779-105115779 on Build GRCh38
rs777632841 Chr.6: 105115732-105115732 on Build GRCh38
rs895600763 Chr.6: 105115717-105115717 on Build GRCh38
rs978704499 Chr.6: 105115830-105115830 on Build GRCh38
rs752335643 Chr.6: 105115816-105115816 on Build GRCh38
rs377316118 Chr.6: 105115685-105115685 on Build GRCh38
rs751114174 Chr.6: 105115698-105115698 on Build GRCh38
rs1029060322 Chr.6: 105115838-105115838 on Build GRCh38
rs765759125 Chr.6: 105115688-105115688 on Build GRCh38
rs147151643 Chr.6: 105115854-105115854 on Build GRCh38
rs541077631 Chr.6: 105115821-105115821 on Build GRCh38
rs931712021 Chr.6: 105115705-105115706 on Build GRCh38

View Full Product Details