Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 43803619-43803892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLVRA
Entrez Gene ID
644
Gene Name
biliverdin reductase A
Gene Aliases
BLVR, BVR, BVRA
UniGene
Hs.488143
Gene Chromosome Location
Chr.7: 43758153-43807342 on Build GRCh38
Transcripts
NM_000712.3 NM_001253823.1 XM_011515474.2 XM_017012520.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6044082 c.481C>T p.R161W Chr.7: 43803696-43803696 on Build GRCh38
746841 c.504C>T p.S168S Chr.7: 43803719-43803719 on Build GRCh38
7357569 c.475G>A p.E159K Chr.7: 43803690-43803690 on Build GRCh38
4665795 c.505G>A p.G169S Chr.7: 43803720-43803720 on Build GRCh38
3832809 c.512C>A p.S171Y Chr.7: 43803727-43803727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767396103 Chr.7: 43803742-43803742 on Build GRCh38
rs776565902 Chr.7: 43803821-43803821 on Build GRCh38
rs756732049 Chr.7: 43803839-43803839 on Build GRCh38
rs773255338 Chr.7: 43803708-43803708 on Build GRCh38
rs777241815 Chr.7: 43803750-43803750 on Build GRCh38
rs1131372 Chr.7: 43803824-43803824 on Build GRCh38
rs1029904131 Chr.7: 43803662-43803662 on Build GRCh38
rs748363460 Chr.7: 43803792-43803792 on Build GRCh38
rs750251555 Chr.7: 43803823-43803823 on Build GRCh38
rs770789238 Chr.7: 43803820-43803820 on Build GRCh38
rs566565833 Chr.7: 43803727-43803727 on Build GRCh38
rs143532590 Chr.7: 43803768-43803768 on Build GRCh38
rs772321543 Chr.7: 43803793-43803793 on Build GRCh38
rs762561277 Chr.7: 43803831-43803831 on Build GRCh38
rs1016829826 Chr.7: 43803720-43803720 on Build GRCh38
rs780247025 Chr.7: 43803666-43803666 on Build GRCh38
rs371055504 Chr.7: 43803801-43803801 on Build GRCh38
rs759355905 Chr.7: 43803822-43803822 on Build GRCh38
rs759271732 Chr.7: 43803723-43803723 on Build GRCh38
rs765437460 Chr.7: 43803660-43803662 on Build GRCh38
rs915952334 Chr.7: 43803783-43803783 on Build GRCh38
rs374145853 Chr.7: 43803683-43803683 on Build GRCh38
rs146795036 Chr.7: 43803697-43803697 on Build GRCh38
rs148879248 Chr.7: 43803749-43803749 on Build GRCh38
rs750976233 Chr.7: 43803755-43803755 on Build GRCh38
rs773161165 Chr.7: 43803798-43803798 on Build GRCh38
rs752396295 Chr.7: 43803825-43803825 on Build GRCh38
rs755445570 Chr.7: 43803866-43803866 on Build GRCh38
rs751016176 Chr.7: 43803837-43803837 on Build GRCh38
rs200781764 Chr.7: 43803682-43803682 on Build GRCh38
rs746311162 Chr.7: 43803652-43803652 on Build GRCh38
rs752906690 Chr.7: 43803835-43803836 on Build GRCh38
rs768984886 Chr.7: 43803688-43803688 on Build GRCh38
rs779302259 Chr.7: 43803781-43803781 on Build GRCh38
rs377670975 Chr.7: 43803696-43803696 on Build GRCh38
rs760722927 Chr.7: 43803804-43803804 on Build GRCh38
rs760502072 Chr.7: 43803713-43803713 on Build GRCh38
rs766797938 Chr.7: 43803855-43803855 on Build GRCh38
rs766264913 Chr.7: 43803717-43803717 on Build GRCh38
rs896000702 Chr.7: 43803740-43803740 on Build GRCh38
rs780523342 Chr.7: 43803758-43803758 on Build GRCh38
rs752304652 Chr.7: 43803745-43803745 on Build GRCh38
rs772231652 Chr.7: 43803707-43803707 on Build GRCh38
rs763723325 Chr.7: 43803834-43803834 on Build GRCh38
rs146563888 Chr.7: 43803719-43803719 on Build GRCh38
rs756570309 Chr.7: 43803757-43803757 on Build GRCh38
rs950169363 Chr.7: 43803807-43803807 on Build GRCh38
rs770242214 Chr.7: 43803660-43803660 on Build GRCh38
rs867045095 Chr.7: 43803775-43803775 on Build GRCh38
rs769105616 Chr.7: 43803771-43803771 on Build GRCh38
rs754351065 Chr.7: 43803859-43803859 on Build GRCh38

View Full Product Details