Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50309041-50309314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB3
Entrez Gene ID
85417
Gene Name
cyclin B3
Gene Aliases
-
UniGene
Hs.130310
Gene Chromosome Location
Chr.X: 50202713-50351914 on Build GRCh38
Transcripts
NM_033031.2 NM_033670.2 XM_017029913.1 XM_017029914.1 XM_017029915.1 XM_017029916.1 XM_017029917.1 XM_017029918.1

SNP Detail

SNP ID Chromosome Location
rs781942029 Chr.X: 50309163-50309163 on Build GRCh38
rs782482934 Chr.X: 50309268-50309268 on Build GRCh38
rs782080579 Chr.X: 50309079-50309079 on Build GRCh38
rs201480633 Chr.X: 50309267-50309267 on Build GRCh38
rs782552876 Chr.X: 50309255-50309255 on Build GRCh38
rs782768940 Chr.X: 50309235-50309235 on Build GRCh38
rs782570615 Chr.X: 50309266-50309266 on Build GRCh38
rs782000483 Chr.X: 50309080-50309080 on Build GRCh38
rs781824721 Chr.X: 50309185-50309185 on Build GRCh38
rs985767020 Chr.X: 50309236-50309236 on Build GRCh38
rs782412756 Chr.X: 50309137-50309137 on Build GRCh38
rs782597059 Chr.X: 50309282-50309282 on Build GRCh38
rs781813119 Chr.X: 50309095-50309095 on Build GRCh38
rs782109179 Chr.X: 50309125-50309125 on Build GRCh38
rs781947698 Chr.X: 50309114-50309114 on Build GRCh38
rs781906156 Chr.X: 50309253-50309253 on Build GRCh38
rs112896746 Chr.X: 50309160-50309160 on Build GRCh38
rs372034679 Chr.X: 50309237-50309237 on Build GRCh38
rs782140683 Chr.X: 50309211-50309211 on Build GRCh38
rs368033012 Chr.X: 50309165-50309165 on Build GRCh38

View Full Product Details