Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 181549529-181549764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERKL
Entrez Gene ID
375298
Gene Name
ceramide kinase like
Gene Aliases
RP26
UniGene
Hs.732358
Gene Chromosome Location
Chr.2: 181536672-181657107 on Build GRCh38
Transcripts
NM_001030311.2 NM_001030312.2 NM_001030313.2 NM_001160277.1 NM_201548.4 NR_027689.1 NR_027690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5599290 c.937C>T p.P313S Chr.2: 181549670-181549670 on Build GRCh38
5625652 c.964A>T p.I322F Chr.2: 181549643-181549643 on Build GRCh38
5599291 c.859C>T p.P287S Chr.2: 181549670-181549670 on Build GRCh38
3896135 c.925C>T p.L309F Chr.2: 181549682-181549682 on Build GRCh38
4893806 c.867G>A p.V289V Chr.2: 181549662-181549662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552603194 Chr.2: 181549613-181549613 on Build GRCh38
rs772798578 Chr.2: 181549681-181549681 on Build GRCh38
rs779802137 Chr.2: 181549637-181549637 on Build GRCh38
rs568190450 Chr.2: 181549703-181549703 on Build GRCh38
rs144831855 Chr.2: 181549671-181549671 on Build GRCh38
rs200369970 Chr.2: 181549699-181549699 on Build GRCh38
rs746847914 Chr.2: 181549655-181549655 on Build GRCh38
rs768174254 Chr.2: 181549698-181549698 on Build GRCh38
rs955229925 Chr.2: 181549647-181549647 on Build GRCh38
rs990281755 Chr.2: 181549695-181549695 on Build GRCh38
rs370924180 Chr.2: 181549611-181549611 on Build GRCh38
rs865824275 Chr.2: 181549560-181549560 on Build GRCh38
rs187829374 Chr.2: 181549621-181549621 on Build GRCh38
rs764481152 Chr.2: 181549597-181549597 on Build GRCh38
rs753943349 Chr.2: 181549609-181549609 on Build GRCh38
rs376192395 Chr.2: 181549582-181549582 on Build GRCh38
rs548192967 Chr.2: 181549669-181549669 on Build GRCh38
rs750564727 Chr.2: 181549708-181549708 on Build GRCh38
rs756369188 Chr.2: 181549584-181549584 on Build GRCh38
rs897797594 Chr.2: 181549586-181549586 on Build GRCh38
rs957155100 Chr.2: 181549683-181549683 on Build GRCh38
rs12623687 Chr.2: 181549631-181549631 on Build GRCh38
rs140777292 Chr.2: 181549665-181549665 on Build GRCh38
rs768283864 Chr.2: 181549660-181549660 on Build GRCh38
rs1014274399 Chr.2: 181549567-181549567 on Build GRCh38
rs753157290 Chr.2: 181549583-181549583 on Build GRCh38
rs759935871 Chr.2: 181549684-181549684 on Build GRCh38
rs769145827 Chr.2: 181549673-181549673 on Build GRCh38
rs370549271 Chr.2: 181549732-181549732 on Build GRCh38
rs765183381 Chr.2: 181549705-181549705 on Build GRCh38
rs764571052 Chr.2: 181549700-181549700 on Build GRCh38

View Full Product Details