Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 235740870-235741143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGAP1
Entrez Gene ID
116987
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 1
Gene Aliases
AGAP-1, CENTG2, GGAP1, cnt-g2
UniGene
Hs.435039
Gene Chromosome Location
Chr.2: 235494089-236131800 on Build GRCh38
Transcripts
NM_001037131.2 NM_001244888.1 NM_014914.4 XM_005246059.4 XM_006712234.3 XM_006712235.3 XM_006712237.3 XM_006712239.3 XM_011510547.2 XM_011510548.2 XM_011510549.2 XM_017003282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
127272 c.381C>G p.G127G Chr.2: 235741033-235741033 on Build GRCh38
1180904 c.381delC p.P129fs*22 Chr.2: 235741033-235741033 on Build GRCh38
1018228 c.311-1G>T p.? Chr.2: 235740962-235740962 on Build GRCh38
1018229 c.336C>T p.V112V Chr.2: 235740988-235740988 on Build GRCh38
7143530 c.355C>T p.L119L Chr.2: 235741007-235741007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774722090 Chr.2: 235740915-235740915 on Build GRCh38
rs758316652 Chr.2: 235741062-235741062 on Build GRCh38
rs369456229 Chr.2: 235741033-235741033 on Build GRCh38
rs373845612 Chr.2: 235741096-235741096 on Build GRCh38
rs751385813 Chr.2: 235740923-235740925 on Build GRCh38
rs997740420 Chr.2: 235741015-235741015 on Build GRCh38
rs371599363 Chr.2: 235741009-235741009 on Build GRCh38
rs190673072 Chr.2: 235740997-235740997 on Build GRCh38
rs748670548 Chr.2: 235741098-235741098 on Build GRCh38
rs756920826 Chr.2: 235740993-235740993 on Build GRCh38
rs140458133 Chr.2: 235741045-235741045 on Build GRCh38
rs767538224 Chr.2: 235741077-235741078 on Build GRCh38
rs146396295 Chr.2: 235740894-235740894 on Build GRCh38
rs534147616 Chr.2: 235741056-235741056 on Build GRCh38
rs753850890 Chr.2: 235741087-235741088 on Build GRCh38
rs759271722 Chr.2: 235741044-235741044 on Build GRCh38
rs773218170 Chr.2: 235741032-235741032 on Build GRCh38
rs773482722 Chr.2: 235740933-235740933 on Build GRCh38
rs139740637 Chr.2: 235740913-235740913 on Build GRCh38
rs887401236 Chr.2: 235740916-235740916 on Build GRCh38
rs764113854 Chr.2: 235741067-235741067 on Build GRCh38
rs59103347 Chr.2: 235741051-235741051 on Build GRCh38
rs185568896 Chr.2: 235740987-235740987 on Build GRCh38
rs761892788 Chr.2: 235740920-235740920 on Build GRCh38
rs754838977 Chr.2: 235740949-235740950 on Build GRCh38
rs182854140 Chr.2: 235741039-235741039 on Build GRCh38
rs753581031 Chr.2: 235740964-235740964 on Build GRCh38
rs568669809 Chr.2: 235740896-235740896 on Build GRCh38
rs373605673 Chr.2: 235741116-235741116 on Build GRCh38
rs756828738 Chr.2: 235741078-235741078 on Build GRCh38
rs757882352 Chr.2: 235740989-235740989 on Build GRCh38
rs181342958 Chr.2: 235740932-235740932 on Build GRCh38
rs769242532 Chr.2: 235740914-235740914 on Build GRCh38
rs754799559 Chr.2: 235740984-235740984 on Build GRCh38
rs968019743 Chr.2: 235741084-235741085 on Build GRCh38
rs752647506 Chr.2: 235741052-235741052 on Build GRCh38
rs144424571 Chr.2: 235741027-235741027 on Build GRCh38
rs780931134 Chr.2: 235741084-235741084 on Build GRCh38
rs760931520 Chr.2: 235740940-235740940 on Build GRCh38
rs772414008 Chr.2: 235741099-235741099 on Build GRCh38
rs1044759265 Chr.2: 235740899-235740899 on Build GRCh38
rs370716770 Chr.2: 235740944-235740944 on Build GRCh38
rs751070159 Chr.2: 235741070-235741070 on Build GRCh38
rs371488003 Chr.2: 235741091-235741091 on Build GRCh38
rs749594933 Chr.2: 235741002-235741002 on Build GRCh38
rs956920805 Chr.2: 235741102-235741102 on Build GRCh38
rs556163643 Chr.2: 235740988-235740988 on Build GRCh38
rs376857250 Chr.2: 235741005-235741005 on Build GRCh38

View Full Product Details