Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 34033251-34033518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF3
Entrez Gene ID
114899
Gene Name
C1q and tumor necrosis factor related protein 3
Gene Aliases
C1ATNF3, CORCS, CORS, CORS-26, CORS26, CTRP3
UniGene
Hs.171929
Gene Chromosome Location
Chr.5: 34017858-34043266 on Build GRCh38
Transcripts
NM_030945.3 NM_181435.5 NR_037951.1

Gene Symbol
C1QTNF3-AMACR
Entrez Gene ID
100534612
Gene Name
C1QTNF3-AMACR readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.171929
Gene Chromosome Location
Chr.5: 33986986-34124528 on Build GRCh38
Transcripts
NM_030945.3 NM_181435.5 NR_037951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1695554 c.556C>T p.P186S Chr.5: 34033318-34033318 on Build GRCh38
3661790 c.539A>G p.K180R Chr.5: 34033335-34033335 on Build GRCh38
5187724 c.570+2T>C p.? Chr.5: 34033302-34033302 on Build GRCh38
5133754 c.550G>A p.G184R Chr.5: 34033324-34033324 on Build GRCh38
211420 c.500C>T p.P167L Chr.5: 34033374-34033374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189735720 Chr.5: 34033361-34033361 on Build GRCh38
rs1009985288 Chr.5: 34033434-34033434 on Build GRCh38
rs760865065 Chr.5: 34033346-34033346 on Build GRCh38
rs377672801 Chr.5: 34033325-34033325 on Build GRCh38
rs768620544 Chr.5: 34033445-34033445 on Build GRCh38
rs778708836 Chr.5: 34033277-34033277 on Build GRCh38
rs749754832 Chr.5: 34033314-34033314 on Build GRCh38
rs370798197 Chr.5: 34033291-34033291 on Build GRCh38
rs749766233 Chr.5: 34033393-34033393 on Build GRCh38
rs772976267 Chr.5: 34033358-34033358 on Build GRCh38
rs1012509142 Chr.5: 34033286-34033286 on Build GRCh38
rs375154760 Chr.5: 34033307-34033307 on Build GRCh38
rs779601069 Chr.5: 34033293-34033293 on Build GRCh38
rs775751421 Chr.5: 34033339-34033339 on Build GRCh38
rs765947533 Chr.5: 34033486-34033486 on Build GRCh38
rs764181192 Chr.5: 34033354-34033354 on Build GRCh38
rs779468479 Chr.5: 34033400-34033400 on Build GRCh38
rs771216364 Chr.5: 34033317-34033317 on Build GRCh38
rs201831968 Chr.5: 34033371-34033371 on Build GRCh38
rs766113889 Chr.5: 34033492-34033492 on Build GRCh38
rs775873735 Chr.5: 34033436-34033436 on Build GRCh38
rs772400068 Chr.5: 34033326-34033326 on Build GRCh38
rs977235537 Chr.5: 34033281-34033281 on Build GRCh38
rs753729578 Chr.5: 34033355-34033355 on Build GRCh38
rs754470860 Chr.5: 34033372-34033372 on Build GRCh38
rs774770764 Chr.5: 34033319-34033319 on Build GRCh38
rs773035202 Chr.5: 34033480-34033480 on Build GRCh38
rs762573935 Chr.5: 34033482-34033483 on Build GRCh38
rs761741428 Chr.5: 34033358-34033358 on Build GRCh38
rs147942281 Chr.5: 34033394-34033394 on Build GRCh38
rs765202151 Chr.5: 34033469-34033469 on Build GRCh38
rs772524434 Chr.5: 34033429-34033429 on Build GRCh38
rs772621983 Chr.5: 34033290-34033290 on Build GRCh38
rs761717549 Chr.5: 34033459-34033459 on Build GRCh38
rs375595390 Chr.5: 34033418-34033418 on Build GRCh38
rs778175026 Chr.5: 34033310-34033310 on Build GRCh38
rs750208893 Chr.5: 34033362-34033362 on Build GRCh38
rs779654581 Chr.5: 34033364-34033364 on Build GRCh38
rs368528853 Chr.5: 34033483-34033483 on Build GRCh38
rs370689464 Chr.5: 34033391-34033391 on Build GRCh38
rs759970201 Chr.5: 34033451-34033451 on Build GRCh38
rs760918499 Chr.5: 34033443-34033443 on Build GRCh38

View Full Product Details