Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101100896-101101141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPI
Entrez Gene ID
2491
Gene Name
centromere protein I
Gene Aliases
CENP-I, FSHPRH1, LRPR1
UniGene
Hs.348920
Gene Chromosome Location
Chr.X: 101098159-101181847 on Build GRCh38
Transcripts
NM_001318521.1 NM_001318523.1 NM_006733.3 XM_005262111.2 XM_011530894.2 XM_011530895.2 XM_011530897.2 XM_011530899.2 XM_017029383.1

SNP Detail

SNP ID Chromosome Location
rs756631764 Chr.X: 101101063-101101063 on Build GRCh38
rs200618447 Chr.X: 101101032-101101036 on Build GRCh38
rs928832756 Chr.X: 101100990-101100990 on Build GRCh38
rs763890747 Chr.X: 101100987-101100987 on Build GRCh38
rs377491282 Chr.X: 101101037-101101041 on Build GRCh38
rs376026355 Chr.X: 101101103-101101103 on Build GRCh38
rs189249919 Chr.X: 101101098-101101098 on Build GRCh38
rs769077146 Chr.X: 101101112-101101112 on Build GRCh38
rs1052869273 Chr.X: 101100998-101100998 on Build GRCh38
rs753934526 Chr.X: 101100937-101100947 on Build GRCh38
rs757368380 Chr.X: 101101027-101101027 on Build GRCh38
rs757364362 Chr.X: 101100956-101100956 on Build GRCh38
rs200931377 Chr.X: 101101033-101101037 on Build GRCh38
rs749116569 Chr.X: 101101004-101101005 on Build GRCh38
rs750487547 Chr.X: 101101032-101101033 on Build GRCh38
rs750665828 Chr.X: 101101036-101101036 on Build GRCh38
rs184304408 Chr.X: 101101085-101101085 on Build GRCh38
rs202234726 Chr.X: 101101034-101101035 on Build GRCh38
rs750489025 Chr.X: 101101043-101101043 on Build GRCh38
rs373474057 Chr.X: 101101088-101101088 on Build GRCh38
rs946314073 Chr.X: 101100946-101100946 on Build GRCh38
rs938476653 Chr.X: 101100963-101100963 on Build GRCh38

View Full Product Details