Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42730426-42730671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC13
Entrez Gene ID
152206
Gene Name
coiled-coil domain containing 13
Gene Aliases
-
UniGene
Hs.604182
Gene Chromosome Location
Chr.3: 42704118-42773253 on Build GRCh38
Transcripts
NM_144719.3 XM_005264898.3 XM_011533418.2 XM_011533419.2 XM_017005781.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3781817 c.1718G>A p.R573Q Chr.3: 42730467-42730467 on Build GRCh38
1044780 c.1717C>T p.R573W Chr.3: 42730468-42730468 on Build GRCh38
7271771 c.1596-9T>G p.? Chr.3: 42730598-42730598 on Build GRCh38
1423135 c.1654C>T p.L552F Chr.3: 42730531-42730531 on Build GRCh38
6213201 c.1668C>T p.A556A Chr.3: 42730517-42730517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764948592 Chr.3: 42730525-42730525 on Build GRCh38
rs781211064 Chr.3: 42730477-42730477 on Build GRCh38
rs777828338 Chr.3: 42730498-42730498 on Build GRCh38
rs752399047 Chr.3: 42730450-42730450 on Build GRCh38
rs545125934 Chr.3: 42730446-42730446 on Build GRCh38
rs530616333 Chr.3: 42730517-42730517 on Build GRCh38
rs370757052 Chr.3: 42730507-42730507 on Build GRCh38
rs775010616 Chr.3: 42730504-42730504 on Build GRCh38
rs771736654 Chr.3: 42730509-42730509 on Build GRCh38
rs868583428 Chr.3: 42730579-42730579 on Build GRCh38
rs760077000 Chr.3: 42730511-42730511 on Build GRCh38
rs998098639 Chr.3: 42730552-42730552 on Build GRCh38
rs201866897 Chr.3: 42730614-42730614 on Build GRCh38
rs942286678 Chr.3: 42730478-42730478 on Build GRCh38
rs147483095 Chr.3: 42730627-42730627 on Build GRCh38
rs776352803 Chr.3: 42730613-42730613 on Build GRCh38
rs765039820 Chr.3: 42730615-42730615 on Build GRCh38
rs914100427 Chr.3: 42730513-42730513 on Build GRCh38
rs750392856 Chr.3: 42730534-42730534 on Build GRCh38
rs561099740 Chr.3: 42730499-42730499 on Build GRCh38
rs558622736 Chr.3: 42730457-42730457 on Build GRCh38
rs779944944 Chr.3: 42730542-42730542 on Build GRCh38
rs139553437 Chr.3: 42730581-42730581 on Build GRCh38
rs370101560 Chr.3: 42730617-42730617 on Build GRCh38
rs368080694 Chr.3: 42730583-42730583 on Build GRCh38
rs969682374 Chr.3: 42730630-42730630 on Build GRCh38
rs753423577 Chr.3: 42730518-42730518 on Build GRCh38
rs778114249 Chr.3: 42730555-42730555 on Build GRCh38
rs138907128 Chr.3: 42730500-42730500 on Build GRCh38
rs760333077 Chr.3: 42730452-42730452 on Build GRCh38
rs756143025 Chr.3: 42730547-42730547 on Build GRCh38
rs199605259 Chr.3: 42730501-42730501 on Build GRCh38
rs775013809 Chr.3: 42730510-42730510 on Build GRCh38
rs943382408 Chr.3: 42730577-42730577 on Build GRCh38
rs749410782 Chr.3: 42730563-42730563 on Build GRCh38
rs373939044 Chr.3: 42730580-42730580 on Build GRCh38
rs753712253 Chr.3: 42730453-42730453 on Build GRCh38
rs200289473 Chr.3: 42730468-42730468 on Build GRCh38
rs12495805 Chr.3: 42730546-42730546 on Build GRCh38
rs141485184 Chr.3: 42730467-42730467 on Build GRCh38
rs763844733 Chr.3: 42730516-42730516 on Build GRCh38
rs532752386 Chr.3: 42730644-42730644 on Build GRCh38
rs763112794 Chr.3: 42730584-42730584 on Build GRCh38
rs778883786 Chr.3: 42730462-42730462 on Build GRCh38
rs761612324 Chr.3: 42730522-42730522 on Build GRCh38
rs758285106 Chr.3: 42730537-42730537 on Build GRCh38
rs1000459528 Chr.3: 42730645-42730645 on Build GRCh38
rs760294365 Chr.3: 42730600-42730600 on Build GRCh38
rs764126478 Chr.3: 42730589-42730589 on Build GRCh38
rs6802779 Chr.3: 42730628-42730628 on Build GRCh38
rs770104913 Chr.3: 42730493-42730493 on Build GRCh38
rs544603189 Chr.3: 42730490-42730490 on Build GRCh38
rs767997400 Chr.3: 42730605-42730605 on Build GRCh38

View Full Product Details