Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36070339-36070570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBR1
Entrez Gene ID
873
Gene Name
carbonyl reductase 1
Gene Aliases
CBR, SDR21C1, hCBR1
UniGene
Hs.88778
Gene Chromosome Location
Chr.21: 36069924-36073177 on Build GRCh38
Transcripts
NM_001286789.1 NM_001757.3 NR_040084.1

Gene Symbol
LOC100133286
Entrez Gene ID
100133286
Gene Name
uncharacterized LOC100133286
Gene Aliases
-
UniGene
Hs.728809
Gene Chromosome Location
Chr.21: 36069642-36126640 on Build GRCh38
Transcripts
NM_001286789.1 NM_001757.3 NR_040084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3389990 c.267C>T p.N89N Chr.21: 36070382-36070382 on Build GRCh38
7220713 c.273G>A p.A91A Chr.21: 36070388-36070388 on Build GRCh38
2844382 c.266A>G p.N89S Chr.21: 36070381-36070381 on Build GRCh38
6585990 c.253G>A p.D85N Chr.21: 36070368-36070368 on Build GRCh38
3405382 c.233G>A p.R78H Chr.21: 36070348-36070348 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748473499 Chr.21: 36070440-36070440 on Build GRCh38
rs780517742 Chr.21: 36070426-36070426 on Build GRCh38
rs1003256032 Chr.21: 36070369-36070369 on Build GRCh38
rs151043730 Chr.21: 36070360-36070360 on Build GRCh38
rs930703290 Chr.21: 36070522-36070522 on Build GRCh38
rs759475325 Chr.21: 36070453-36070453 on Build GRCh38
rs993484656 Chr.21: 36070484-36070484 on Build GRCh38
rs538650389 Chr.21: 36070462-36070462 on Build GRCh38
rs776688059 Chr.21: 36070381-36070381 on Build GRCh38
rs759503884 Chr.21: 36070539-36070539 on Build GRCh38
rs747591615 Chr.21: 36070427-36070427 on Build GRCh38
rs374191813 Chr.21: 36070411-36070411 on Build GRCh38
rs766413880 Chr.21: 36070391-36070391 on Build GRCh38
rs1143663 Chr.21: 36070377-36070377 on Build GRCh38
rs74484583 Chr.21: 36070405-36070405 on Build GRCh38
rs763111396 Chr.21: 36070446-36070446 on Build GRCh38
rs980614462 Chr.21: 36070544-36070544 on Build GRCh38
rs756821807 Chr.21: 36070418-36070418 on Build GRCh38
rs754702330 Chr.21: 36070403-36070403 on Build GRCh38
rs200615385 Chr.21: 36070442-36070442 on Build GRCh38
rs191556963 Chr.21: 36070388-36070388 on Build GRCh38
rs773365505 Chr.21: 36070390-36070390 on Build GRCh38
rs751602193 Chr.21: 36070402-36070402 on Build GRCh38
rs762056579 Chr.21: 36070385-36070385 on Build GRCh38
rs372726650 Chr.21: 36070451-36070451 on Build GRCh38
rs111862586 Chr.21: 36070431-36070431 on Build GRCh38
rs771180650 Chr.21: 36070448-36070448 on Build GRCh38
rs769114657 Chr.21: 36070364-36070364 on Build GRCh38
rs774277249 Chr.21: 36070450-36070450 on Build GRCh38
rs1049555217 Chr.21: 36070522-36070522 on Build GRCh38
rs41540212 Chr.21: 36070429-36070429 on Build GRCh38
rs748300804 Chr.21: 36070453-36070458 on Build GRCh38
rs556896935 Chr.21: 36070467-36070467 on Build GRCh38
rs755404701 Chr.21: 36070486-36070486 on Build GRCh38
rs1030669258 Chr.21: 36070384-36070384 on Build GRCh38
rs767383110 Chr.21: 36070404-36070404 on Build GRCh38
rs951920461 Chr.21: 36070542-36070542 on Build GRCh38
rs1000335693 Chr.21: 36070386-36070386 on Build GRCh38
rs200695117 Chr.21: 36070359-36070359 on Build GRCh38
rs9979957 Chr.21: 36070464-36070464 on Build GRCh38
rs571954075 Chr.21: 36070503-36070503 on Build GRCh38
rs777409910 Chr.21: 36070406-36070406 on Build GRCh38

View Full Product Details