Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75205829-75206102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf39
Entrez Gene ID
56905
Gene Name
chromosome 15 open reading frame 39
Gene Aliases
-
UniGene
Hs.17936
Gene Chromosome Location
Chr.15: 75201880-75212169 on Build GRCh38
Transcripts
NM_015492.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6614408 c.31G>A p.G11R Chr.15: 75206079-75206079 on Build GRCh38
6614407 c.31G>A p.G11R Chr.15: 75206079-75206079 on Build GRCh38
3794409 c.21G>A p.L7L Chr.15: 75206069-75206069 on Build GRCh38
4056948 c.4G>A p.A2T Chr.15: 75206052-75206052 on Build GRCh38
4517112 c.26_27CC>TT p.T9I Chr.15: 75206074-75206075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747678168 Chr.15: 75206077-75206077 on Build GRCh38
rs765358715 Chr.15: 75206007-75206007 on Build GRCh38
rs543040553 Chr.15: 75205915-75205915 on Build GRCh38
rs958806766 Chr.15: 75205950-75205950 on Build GRCh38
rs1015147446 Chr.15: 75205901-75205901 on Build GRCh38
rs544130198 Chr.15: 75206048-75206048 on Build GRCh38
rs370912089 Chr.15: 75206064-75206064 on Build GRCh38
rs751754386 Chr.15: 75206047-75206047 on Build GRCh38
rs146353943 Chr.15: 75205937-75205937 on Build GRCh38
rs545529967 Chr.15: 75205858-75205858 on Build GRCh38
rs1020371547 Chr.15: 75205865-75205866 on Build GRCh38
rs758404364 Chr.15: 75206019-75206019 on Build GRCh38
rs368173977 Chr.15: 75206056-75206056 on Build GRCh38
rs943315860 Chr.15: 75206006-75206006 on Build GRCh38
rs1049390558 Chr.15: 75205979-75205979 on Build GRCh38
rs778866464 Chr.15: 75205880-75205880 on Build GRCh38
rs914974262 Chr.15: 75206069-75206069 on Build GRCh38
rs1033286774 Chr.15: 75205886-75205886 on Build GRCh38
rs139089591 Chr.15: 75205989-75205989 on Build GRCh38
rs75710563 Chr.15: 75206033-75206033 on Build GRCh38
rs564658480 Chr.15: 75206030-75206030 on Build GRCh38
rs930511473 Chr.15: 75205966-75205966 on Build GRCh38
rs374040917 Chr.15: 75206034-75206034 on Build GRCh38
rs991496867 Chr.15: 75205912-75205912 on Build GRCh38
rs780056068 Chr.15: 75206031-75206031 on Build GRCh38
rs967529085 Chr.15: 75205876-75205876 on Build GRCh38
rs1050108890 Chr.15: 75205856-75205856 on Build GRCh38
rs573923541 Chr.15: 75206018-75206018 on Build GRCh38
rs991436850 Chr.15: 75205994-75205994 on Build GRCh38
rs757303738 Chr.15: 75205850-75205850 on Build GRCh38
rs780644935 Chr.15: 75206062-75206062 on Build GRCh38
rs865817368 Chr.15: 75206028-75206028 on Build GRCh38
rs781585751 Chr.15: 75206052-75206052 on Build GRCh38
rs771661306 Chr.15: 75206081-75206081 on Build GRCh38
rs771930954 Chr.15: 75206016-75206016 on Build GRCh38
rs959240291 Chr.15: 75205893-75205893 on Build GRCh38
rs554277025 Chr.15: 75205882-75205882 on Build GRCh38
rs191263398 Chr.15: 75205913-75205913 on Build GRCh38
rs746407054 Chr.15: 75206055-75206055 on Build GRCh38
rs950073114 Chr.15: 75205914-75205914 on Build GRCh38
rs890354012 Chr.15: 75205862-75205862 on Build GRCh38

View Full Product Details