Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65198007-65198232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CILP
Entrez Gene ID
8483
Gene Name
cartilage intermediate layer protein
Gene Aliases
CILP-1, HsT18872
UniGene
Hs.442180
Gene Chromosome Location
Chr.15: 65195999-65211608 on Build GRCh38
Transcripts
NM_003613.3 XM_017022678.1 XM_017022679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1238515 c.2122T>C p.W708R Chr.15: 65198164-65198164 on Build GRCh38
434156 c.2133A>T p.E711D Chr.15: 65198153-65198153 on Build GRCh38
4595988 c.2128G>A p.E710K Chr.15: 65198158-65198158 on Build GRCh38
4056275 c.2209C>T p.R737C Chr.15: 65198077-65198077 on Build GRCh38
336483 c.2251C>A p.R751R Chr.15: 65198035-65198035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943945981 Chr.15: 65198047-65198047 on Build GRCh38
rs768154426 Chr.15: 65198059-65198059 on Build GRCh38
rs995585251 Chr.15: 65198136-65198136 on Build GRCh38
rs758380040 Chr.15: 65198203-65198203 on Build GRCh38
rs757488703 Chr.15: 65198175-65198175 on Build GRCh38
rs776366924 Chr.15: 65198076-65198076 on Build GRCh38
rs754098502 Chr.15: 65198156-65198156 on Build GRCh38
rs150788279 Chr.15: 65198201-65198201 on Build GRCh38
rs764167212 Chr.15: 65198134-65198134 on Build GRCh38
rs772774382 Chr.15: 65198095-65198095 on Build GRCh38
rs769347962 Chr.15: 65198092-65198092 on Build GRCh38
rs779039800 Chr.15: 65198183-65198183 on Build GRCh38
rs562122822 Chr.15: 65198077-65198077 on Build GRCh38
rs201712854 Chr.15: 65198204-65198204 on Build GRCh38
rs745797617 Chr.15: 65198031-65198031 on Build GRCh38
rs142177709 Chr.15: 65198103-65198103 on Build GRCh38
rs762686916 Chr.15: 65198189-65198189 on Build GRCh38
rs778651058 Chr.15: 65198029-65198029 on Build GRCh38
rs773447323 Chr.15: 65198060-65198063 on Build GRCh38
rs77484154 Chr.15: 65198026-65198026 on Build GRCh38
rs768508244 Chr.15: 65198213-65198213 on Build GRCh38
rs201186001 Chr.15: 65198209-65198209 on Build GRCh38
rs139906949 Chr.15: 65198034-65198034 on Build GRCh38
rs761054336 Chr.15: 65198115-65198115 on Build GRCh38
rs775403820 Chr.15: 65198035-65198035 on Build GRCh38

View Full Product Details