Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 126425861-126426122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf90
Entrez Gene ID
118611
Gene Name
chromosome 10 open reading frame 90
Gene Aliases
FATS, bA422P15.2
UniGene
Hs.587663
Gene Chromosome Location
Chr.10: 126424997-126671038 on Build GRCh38
Transcripts
NM_001004298.2 XM_005269491.1 XM_005269492.2 XM_011539216.1 XM_011539217.2 XM_011539220.2 XM_011539222.2 XM_011539223.2 XM_011539224.2 XM_011539226.1 XM_011539227.2 XM_011539230.2 XM_011539231.1 XM_011539232.1 XM_011539233.1 XM_017015634.1

Gene Symbol
LOC728158
Entrez Gene ID
728158
Gene Name
hCG2044975
Gene Aliases
-
UniGene
Hs.646701
Gene Chromosome Location
Chr.10: 126423107-126440535 on Build GRCh38
Transcripts
NM_001004298.2 XM_005269491.1 XM_005269492.2 XM_011539216.1 XM_011539217.2 XM_011539220.2 XM_011539222.2 XM_011539223.2 XM_011539224.2 XM_011539226.1 XM_011539227.2 XM_011539230.2 XM_011539231.1 XM_011539232.1 XM_011539233.1 XM_017015634.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4778459 c.2336C>A p.A779D Chr.10: 126426007-126426007 on Build GRCh38
6065327 c.2022C>T p.I674I Chr.10: 126426030-126426030 on Build GRCh38
4766561 c.2356T>G p.L786V Chr.10: 126425899-126425899 on Build GRCh38
4766560 c.2065T>G p.L689V Chr.10: 126425899-126425899 on Build GRCh38
6502240 c.2283G>A p.K761K Chr.10: 126426060-126426060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374088681 Chr.10: 126426059-126426059 on Build GRCh38
rs777386692 Chr.10: 126426055-126426060 on Build GRCh38
rs1016007416 Chr.10: 126426060-126426060 on Build GRCh38
rs767965306 Chr.10: 126426093-126426093 on Build GRCh38
rs1043882905 Chr.10: 126425975-126425975 on Build GRCh38
rs146332380 Chr.10: 126426071-126426071 on Build GRCh38
rs769373786 Chr.10: 126425920-126425920 on Build GRCh38
rs370672577 Chr.10: 126426010-126426010 on Build GRCh38
rs775678581 Chr.10: 126426073-126426073 on Build GRCh38
rs374265832 Chr.10: 126425979-126425979 on Build GRCh38
rs780671267 Chr.10: 126425906-126425906 on Build GRCh38
rs187441937 Chr.10: 126426094-126426094 on Build GRCh38
rs200783026 Chr.10: 126426072-126426072 on Build GRCh38
rs771505495 Chr.10: 126425891-126425891 on Build GRCh38
rs538443729 Chr.10: 126425929-126425929 on Build GRCh38
rs751810351 Chr.10: 126426083-126426083 on Build GRCh38
rs149388609 Chr.10: 126425991-126425991 on Build GRCh38
rs753912532 Chr.10: 126425962-126425962 on Build GRCh38
rs775944482 Chr.10: 126426080-126426080 on Build GRCh38
rs768316290 Chr.10: 126425936-126425936 on Build GRCh38
rs376334305 Chr.10: 126425941-126425941 on Build GRCh38
rs779592842 Chr.10: 126426024-126426024 on Build GRCh38
rs757743695 Chr.10: 126426006-126426006 on Build GRCh38
rs746273682 Chr.10: 126426061-126426078 on Build GRCh38
rs745436700 Chr.10: 126425908-126425908 on Build GRCh38
rs764430485 Chr.10: 126426081-126426081 on Build GRCh38
rs759691712 Chr.10: 126425968-126425968 on Build GRCh38
rs748864137 Chr.10: 126426030-126426030 on Build GRCh38
rs910402979 Chr.10: 126425917-126425917 on Build GRCh38
rs371057454 Chr.10: 126425973-126425973 on Build GRCh38
rs1002391361 Chr.10: 126425980-126425980 on Build GRCh38
rs768422156 Chr.10: 126426036-126426036 on Build GRCh38
rs752883714 Chr.10: 126425982-126425982 on Build GRCh38
rs774003638 Chr.10: 126426050-126426050 on Build GRCh38
rs182354709 Chr.10: 126425986-126425986 on Build GRCh38
rs772267163 Chr.10: 126425934-126425934 on Build GRCh38
rs892714565 Chr.10: 126425931-126425931 on Build GRCh38
rs769318720 Chr.10: 126426023-126426023 on Build GRCh38
rs1053881009 Chr.10: 126425926-126425926 on Build GRCh38
rs541811427 Chr.10: 126426057-126426057 on Build GRCh38
rs757653481 Chr.10: 126426085-126426085 on Build GRCh38
rs762762976 Chr.10: 126425932-126425932 on Build GRCh38
rs774243651 Chr.10: 126425938-126425938 on Build GRCh38
rs770142843 Chr.10: 126426071-126426071 on Build GRCh38
rs752025882 Chr.10: 126426005-126426005 on Build GRCh38
rs767621530 Chr.10: 126425942-126425942 on Build GRCh38

View Full Product Details