Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 113049634-113049905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANK2
Entrez Gene ID
287
Gene Name
ankyrin 2, neuronal
Gene Aliases
ANK-2, LQT4, brank-2
UniGene
Hs.620557
Gene Chromosome Location
Chr.4: 112706083-113383740 on Build GRCh38
Transcripts
NM_001127493.1 NM_001148.4 NM_020977.3 XM_005262945.2 XM_017008068.1 XM_017008070.1 XM_017008071.1 XM_017008072.1 XM_017008073.1 XM_017008074.1 XM_017008075.1 XM_017008076.1 XM_017008077.1 XM_017008078.1 XM_017008079.1 XM_017008080.1 XM_017008081.1 XM_017008084.1 XM_017008085.1 XM_017008086.1 XM_017008088.1 XM_017008091.1 XM_017008092.1 XM_017008093.1 XM_017008094.1 XM_017008095.1 XM_017008096.1 XM_017008097.1 XM_017008098.1 XM_017008099.1 XM_017008100.1 XM_017008101.1 XM_017008102.1 XM_017008103.1 XM_017008104.1 XM_017008105.1 XM_017008106.1 XM_017008107.1 XM_017008108.1 XM_017008109.1 XM_017008110.1 XM_017008111.1 XM_017008112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6166280 c.64A>G p.R22G Chr.4: 113049792-113049792 on Build GRCh38
731919 c.16G>T p.A6S Chr.4: 113049744-113049744 on Build GRCh38
6166281 c.57T>A p.S19R Chr.4: 113049785-113049785 on Build GRCh38
7332009 c.6G>A p.M2I Chr.4: 113049734-113049734 on Build GRCh38
7253585 c.34A>C p.S12R Chr.4: 113049762-113049762 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766477217 Chr.4: 113049704-113049704 on Build GRCh38
rs758753200 Chr.4: 113049697-113049697 on Build GRCh38
rs928291078 Chr.4: 113049806-113049806 on Build GRCh38
rs369260005 Chr.4: 113049784-113049784 on Build GRCh38
rs139765650 Chr.4: 113049812-113049812 on Build GRCh38
rs767932554 Chr.4: 113049838-113049838 on Build GRCh38
rs929854524 Chr.4: 113049732-113049732 on Build GRCh38
rs769843073 Chr.4: 113049808-113049808 on Build GRCh38
rs756605353 Chr.4: 113049743-113049743 on Build GRCh38
rs973992723 Chr.4: 113049689-113049689 on Build GRCh38
rs553045017 Chr.4: 113049883-113049883 on Build GRCh38
rs778450160 Chr.4: 113049762-113049762 on Build GRCh38
rs754306612 Chr.4: 113049857-113049857 on Build GRCh38
rs776225772 Chr.4: 113049689-113049689 on Build GRCh38
rs757871337 Chr.4: 113049863-113049863 on Build GRCh38
rs755320429 Chr.4: 113049716-113049716 on Build GRCh38
rs753042532 Chr.4: 113049844-113049844 on Build GRCh38
rs771799732 Chr.4: 113049766-113049766 on Build GRCh38
rs376816836 Chr.4: 113049682-113049682 on Build GRCh38
rs368586736 Chr.4: 113049688-113049688 on Build GRCh38
rs147439862 Chr.4: 113049790-113049790 on Build GRCh38
rs766430801 Chr.4: 113049820-113049820 on Build GRCh38
rs779606672 Chr.4: 113049776-113049776 on Build GRCh38
rs986220923 Chr.4: 113049668-113049668 on Build GRCh38
rs372870729 Chr.4: 113049690-113049690 on Build GRCh38
rs535919946 Chr.4: 113049738-113049738 on Build GRCh38
rs959061898 Chr.4: 113049676-113049676 on Build GRCh38
rs983314290 Chr.4: 113049770-113049770 on Build GRCh38
rs748654930 Chr.4: 113049741-113049741 on Build GRCh38
rs372063569 Chr.4: 113049699-113049699 on Build GRCh38
rs759736399 Chr.4: 113049823-113049823 on Build GRCh38
rs769446762 Chr.4: 113049866-113049866 on Build GRCh38
rs201646790 Chr.4: 113049711-113049711 on Build GRCh38
rs745356870 Chr.4: 113049765-113049765 on Build GRCh38
rs746628566 Chr.4: 113049781-113049781 on Build GRCh38
rs188114138 Chr.4: 113049868-113049868 on Build GRCh38
rs756660731 Chr.4: 113049849-113049849 on Build GRCh38
rs778219314 Chr.4: 113049854-113049854 on Build GRCh38
rs770937235 Chr.4: 113049670-113049670 on Build GRCh38
rs762973067 Chr.4: 113049819-113049819 on Build GRCh38
rs768352926 Chr.4: 113049783-113049783 on Build GRCh38
rs765412554 Chr.4: 113049683-113049683 on Build GRCh38

View Full Product Details