Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 93369974-93370237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEUP1
Entrez Gene ID
159989
Gene Name
deuterosome assembly protein 1
Gene Aliases
CCDC67
UniGene
Hs.436625
Gene Chromosome Location
Chr.11: 93329079-93438470 on Build GRCh38
Transcripts
NM_181645.3 XM_005273802.1 XM_011542629.2 XM_011542630.1 XM_011542631.2 XM_011542632.2 XM_011542633.2 XM_011542635.2 XM_011542637.2 XM_017017245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3686044 c.433-2_433-1insC p.? Chr.11: 93370071-93370072 on Build GRCh38
5517837 c.480T>A p.Y160* Chr.11: 93370120-93370120 on Build GRCh38
147306 c.456A>G p.E152E Chr.11: 93370096-93370096 on Build GRCh38
6562554 c.508C>A p.Q170K Chr.11: 93370148-93370148 on Build GRCh38
3454674 c.481C>T p.Q161* Chr.11: 93370121-93370121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044765793 Chr.11: 93370008-93370008 on Build GRCh38
rs757916054 Chr.11: 93370186-93370187 on Build GRCh38
rs768604433 Chr.11: 93370132-93370132 on Build GRCh38
rs754570032 Chr.11: 93370175-93370175 on Build GRCh38
rs1030181697 Chr.11: 93370112-93370112 on Build GRCh38
rs34205920 Chr.11: 93370164-93370164 on Build GRCh38
rs760400259 Chr.11: 93370166-93370166 on Build GRCh38
rs866931740 Chr.11: 93370098-93370098 on Build GRCh38
rs747454592 Chr.11: 93370078-93370078 on Build GRCh38
rs751446069 Chr.11: 93370035-93370035 on Build GRCh38
rs752214057 Chr.11: 93370187-93370187 on Build GRCh38
rs774395193 Chr.11: 93370137-93370137 on Build GRCh38
rs781443960 Chr.11: 93370082-93370082 on Build GRCh38
rs753355639 Chr.11: 93370063-93370063 on Build GRCh38
rs763193371 Chr.11: 93370118-93370118 on Build GRCh38
rs567563769 Chr.11: 93370163-93370163 on Build GRCh38
rs192782337 Chr.11: 93370170-93370170 on Build GRCh38
rs781742562 Chr.11: 93370202-93370202 on Build GRCh38
rs1041429631 Chr.11: 93370007-93370007 on Build GRCh38
rs770156602 Chr.11: 93370101-93370101 on Build GRCh38
rs754414914 Chr.11: 93370068-93370068 on Build GRCh38
rs757728297 Chr.11: 93370189-93370189 on Build GRCh38
rs771370232 Chr.11: 93370081-93370081 on Build GRCh38
rs767193279 Chr.11: 93370046-93370046 on Build GRCh38
rs963460248 Chr.11: 93370106-93370106 on Build GRCh38
rs756580836 Chr.11: 93370045-93370046 on Build GRCh38
rs778258481 Chr.11: 93370184-93370184 on Build GRCh38
rs761725255 Chr.11: 93370042-93370042 on Build GRCh38
rs188249453 Chr.11: 93370055-93370055 on Build GRCh38
rs778298959 Chr.11: 93370156-93370158 on Build GRCh38
rs750114884 Chr.11: 93370047-93370047 on Build GRCh38
rs761466399 Chr.11: 93370178-93370191 on Build GRCh38
rs370253158 Chr.11: 93370173-93370173 on Build GRCh38
rs897684182 Chr.11: 93370074-93370074 on Build GRCh38
rs571383666 Chr.11: 93370204-93370204 on Build GRCh38
rs902846473 Chr.11: 93370009-93370009 on Build GRCh38
rs77230678 Chr.11: 93370115-93370115 on Build GRCh38
rs761623508 Chr.11: 93370159-93370159 on Build GRCh38
rs987835800 Chr.11: 93370125-93370125 on Build GRCh38
rs779631543 Chr.11: 93370062-93370062 on Build GRCh38
rs1003571109 Chr.11: 93370027-93370027 on Build GRCh38
rs530435017 Chr.11: 93370100-93370100 on Build GRCh38

View Full Product Details