Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206116038-206115793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVPR1B
Entrez Gene ID
553
Gene Name
arginine vasopressin receptor 1B
Gene Aliases
AVPR3
UniGene
Hs.1372
Gene Chromosome Location
Chr.1: 206109849-206117048 on Build GRCh38
Transcripts
NM_000707.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4027494 c.918G>A p.W306* Chr.1: 206115973-206115973 on Build GRCh38
1660712 c.862C>A p.L288M Chr.1: 206116029-206116029 on Build GRCh38
2215761 c.873C>T p.I291I Chr.1: 206116018-206116018 on Build GRCh38
3864274 c.889T>A p.F297I Chr.1: 206116002-206116002 on Build GRCh38
1338258 c.865G>A p.A289T Chr.1: 206116026-206116026 on Build GRCh38
View More

View Full Product Details