Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 139042720-139042977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABRACL
Entrez Gene ID
58527
Gene Name
ABRA C-terminal like
Gene Aliases
C6orf115, Costars, HSPC280, PRO2013
UniGene
Hs.600861
Gene Chromosome Location
Chr.6: 139028682-139043302 on Build GRCh38
Transcripts
NM_021243.2 XM_017011151.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7447594 c.218A>G p.D73G Chr.6: 139042875-139042875 on Build GRCh38
5996898 c.159A>G p.A53A Chr.6: 139042816-139042816 on Build GRCh38
5629607 c.78A>T p.L26F Chr.6: 139042735-139042735 on Build GRCh38
7353783 c.106G>T p.D36Y Chr.6: 139042763-139042763 on Build GRCh38
7309943 c.106G>A p.D36N Chr.6: 139042763-139042763 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370701225 Chr.6: 139042903-139042903 on Build GRCh38
rs755564745 Chr.6: 139042865-139042865 on Build GRCh38
rs752173600 Chr.6: 139042756-139042756 on Build GRCh38
rs779344089 Chr.6: 139042760-139042760 on Build GRCh38
rs376913110 Chr.6: 139042863-139042863 on Build GRCh38
rs373650228 Chr.6: 139042915-139042915 on Build GRCh38
rs543813480 Chr.6: 139042926-139042926 on Build GRCh38
rs747243213 Chr.6: 139042793-139042793 on Build GRCh38
rs762945740 Chr.6: 139042823-139042823 on Build GRCh38
rs866833165 Chr.6: 139042800-139042800 on Build GRCh38
rs780732523 Chr.6: 139042754-139042754 on Build GRCh38
rs759227190 Chr.6: 139042853-139042853 on Build GRCh38
rs766084351 Chr.6: 139042763-139042763 on Build GRCh38
rs779848870 Chr.6: 139042936-139042939 on Build GRCh38
rs751142655 Chr.6: 139042781-139042781 on Build GRCh38
rs751241615 Chr.6: 139042744-139042744 on Build GRCh38
rs768476146 Chr.6: 139042877-139042877 on Build GRCh38
rs761932883 Chr.6: 139042817-139042817 on Build GRCh38
rs747355040 Chr.6: 139042913-139042913 on Build GRCh38
rs554883664 Chr.6: 139042748-139042748 on Build GRCh38
rs560335878 Chr.6: 139042764-139042765 on Build GRCh38
rs772960368 Chr.6: 139042821-139042821 on Build GRCh38
rs754412867 Chr.6: 139042849-139042849 on Build GRCh38
rs777259988 Chr.6: 139042868-139042868 on Build GRCh38
rs372618247 Chr.6: 139042820-139042820 on Build GRCh38
rs995722634 Chr.6: 139042775-139042775 on Build GRCh38
rs772384268 Chr.6: 139042776-139042776 on Build GRCh38
rs764431624 Chr.6: 139042807-139042807 on Build GRCh38
rs781503731 Chr.6: 139042925-139042925 on Build GRCh38
rs1039243025 Chr.6: 139042874-139042874 on Build GRCh38
rs754648030 Chr.6: 139042747-139042747 on Build GRCh38
rs751351295 Chr.6: 139042842-139042842 on Build GRCh38
rs573558747 Chr.6: 139042784-139042784 on Build GRCh38
rs200259260 Chr.6: 139042839-139042839 on Build GRCh38
rs766149541 Chr.6: 139042837-139042837 on Build GRCh38
rs941021278 Chr.6: 139042761-139042761 on Build GRCh38
rs201624556 Chr.6: 139042801-139042801 on Build GRCh38
rs767116535 Chr.6: 139042862-139042862 on Build GRCh38

View Full Product Details