Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7368818-7369091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD163L1
Entrez Gene ID
283316
Gene Name
CD163 molecule like 1
Gene Aliases
CD163B, M160, SCARI2
UniGene
Hs.631727
Gene Chromosome Location
Chr.12: 7320510-7444185 on Build GRCh38
Transcripts
NM_001297650.1 NM_174941.5 XM_011520616.1 XM_011520617.2 XM_011520618.2 XM_011520619.1 XM_011520620.1 XM_011520621.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5024231 c.4066T>C p.S1356P Chr.12: 7368939-7368939 on Build GRCh38
139942 c.4067C>T p.S1356F Chr.12: 7368938-7368938 on Build GRCh38
5414821 c.4040-4_4040-3delTC p.? Chr.12: 7368968-7368969 on Build GRCh38
5375986 c.4055C>T p.S1352L Chr.12: 7368950-7368950 on Build GRCh38
431833 c.4042C>A p.Q1348K Chr.12: 7368963-7368963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763563642 Chr.12: 7369019-7369019 on Build GRCh38
rs879031815 Chr.12: 7368979-7368979 on Build GRCh38
rs759431622 Chr.12: 7368958-7368958 on Build GRCh38
rs145256685 Chr.12: 7368939-7368939 on Build GRCh38
rs140872991 Chr.12: 7369045-7369045 on Build GRCh38
rs749743782 Chr.12: 7368975-7368976 on Build GRCh38
rs148052421 Chr.12: 7369012-7369012 on Build GRCh38
rs752036295 Chr.12: 7368891-7368891 on Build GRCh38
rs959092677 Chr.12: 7368886-7368886 on Build GRCh38
rs781647124 Chr.12: 7368899-7368899 on Build GRCh38
rs755083069 Chr.12: 7369039-7369039 on Build GRCh38
rs868394273 Chr.12: 7368966-7368966 on Build GRCh38
rs758628112 Chr.12: 7368998-7368998 on Build GRCh38
rs775301901 Chr.12: 7368968-7368968 on Build GRCh38
rs771878446 Chr.12: 7368959-7368959 on Build GRCh38
rs116763406 Chr.12: 7368908-7368908 on Build GRCh38
rs749386143 Chr.12: 7368930-7368930 on Build GRCh38
rs902064133 Chr.12: 7369016-7369016 on Build GRCh38
rs996787056 Chr.12: 7369034-7369034 on Build GRCh38
rs1003307164 Chr.12: 7368880-7368880 on Build GRCh38
rs761233165 Chr.12: 7368990-7368990 on Build GRCh38
rs148414953 Chr.12: 7368968-7368969 on Build GRCh38
rs1042668180 Chr.12: 7369046-7369046 on Build GRCh38
rs777738943 Chr.12: 7368979-7368979 on Build GRCh38
rs772728915 Chr.12: 7368948-7368948 on Build GRCh38
rs759485909 Chr.12: 7368989-7368990 on Build GRCh38
rs781351459 Chr.12: 7368967-7368968 on Build GRCh38
rs775062285 Chr.12: 7369052-7369052 on Build GRCh38
rs1033457378 Chr.12: 7368883-7368883 on Build GRCh38
rs750604728 Chr.12: 7368982-7368982 on Build GRCh38
rs760219214 Chr.12: 7368979-7368981 on Build GRCh38
rs773775330 Chr.12: 7368968-7368975 on Build GRCh38
rs752027505 Chr.12: 7368993-7368993 on Build GRCh38
rs753546717 Chr.12: 7369061-7369061 on Build GRCh38
rs760455518 Chr.12: 7368977-7368977 on Build GRCh38
rs372157792 Chr.12: 7368949-7368949 on Build GRCh38
rs773550712 Chr.12: 7369038-7369038 on Build GRCh38
rs553606389 Chr.12: 7368992-7368992 on Build GRCh38
rs758320741 Chr.12: 7368977-7368977 on Build GRCh38
rs751985078 Chr.12: 7369040-7369040 on Build GRCh38
rs267603672 Chr.12: 7368950-7368950 on Build GRCh38
rs761647734 Chr.12: 7368897-7368897 on Build GRCh38
rs776289623 Chr.12: 7368987-7368987 on Build GRCh38
rs201580844 Chr.12: 7369002-7369002 on Build GRCh38
rs111404478 Chr.12: 7368873-7368873 on Build GRCh38
rs770990028 Chr.12: 7368932-7368932 on Build GRCh38
rs763857704 Chr.12: 7368999-7369008 on Build GRCh38
rs756092467 Chr.12: 7368969-7368970 on Build GRCh38
rs112121784 Chr.12: 7368929-7368929 on Build GRCh38
rs770181475 Chr.12: 7368910-7368910 on Build GRCh38
rs749535745 Chr.12: 7368968-7368971 on Build GRCh38
rs1038999817 Chr.12: 7368984-7368984 on Build GRCh38
rs59576617 Chr.12: 7368991-7368992 on Build GRCh38
rs778052727 Chr.12: 7368916-7368916 on Build GRCh38
rs780438994 Chr.12: 7368890-7368890 on Build GRCh38

View Full Product Details