Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 58516941-58517214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPK2
Entrez Gene ID
115701
Gene Name
alpha kinase 2
Gene Aliases
HAK
UniGene
Hs.628152
Gene Chromosome Location
Chr.18: 58481247-58629192 on Build GRCh38
Transcripts
NM_052947.3 XM_011525801.2 XM_011525802.2 XM_017025535.1 XM_017025536.1 XM_017025537.1 XM_017025538.1 XM_017025539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
989243 c.3821G>A p.R1274H Chr.18: 58517110-58517110 on Build GRCh38
259396 c.5707G>T p.D1903Y Chr.18: 58517141-58517141 on Build GRCh38
3526712 c.5754C>T p.I1918I Chr.18: 58517094-58517094 on Build GRCh38
5607272 c.5799C>T p.A1933A Chr.18: 58517049-58517049 on Build GRCh38
989239 c.3969T>C p.Y1323Y Chr.18: 58516962-58516962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754653638 Chr.18: 58517105-58517105 on Build GRCh38
rs55647142 Chr.18: 58517065-58517065 on Build GRCh38
rs768830207 Chr.18: 58517162-58517162 on Build GRCh38
rs753524863 Chr.18: 58517044-58517044 on Build GRCh38
rs763131723 Chr.18: 58517078-58517078 on Build GRCh38
rs56249361 Chr.18: 58516980-58516980 on Build GRCh38
rs1004905892 Chr.18: 58516996-58516996 on Build GRCh38
rs778586800 Chr.18: 58517110-58517110 on Build GRCh38
rs886411789 Chr.18: 58516970-58516970 on Build GRCh38
rs374271622 Chr.18: 58517093-58517093 on Build GRCh38
rs748143041 Chr.18: 58517047-58517047 on Build GRCh38
rs3809984 Chr.18: 58517045-58517045 on Build GRCh38
rs757741609 Chr.18: 58517006-58517006 on Build GRCh38
rs746962671 Chr.18: 58517140-58517140 on Build GRCh38
rs774423134 Chr.18: 58517166-58517166 on Build GRCh38
rs752104140 Chr.18: 58517005-58517005 on Build GRCh38
rs777629986 Chr.18: 58517123-58517123 on Build GRCh38
rs375621243 Chr.18: 58516973-58516973 on Build GRCh38
rs34304827 Chr.18: 58517172-58517172 on Build GRCh38
rs1049182102 Chr.18: 58516969-58516969 on Build GRCh38
rs927412152 Chr.18: 58517114-58517114 on Build GRCh38
rs754205739 Chr.18: 58517193-58517193 on Build GRCh38
rs372914995 Chr.18: 58517089-58517089 on Build GRCh38
rs151328897 Chr.18: 58517087-58517087 on Build GRCh38
rs967856650 Chr.18: 58517058-58517058 on Build GRCh38
rs770674676 Chr.18: 58517141-58517141 on Build GRCh38
rs867597033 Chr.18: 58517157-58517157 on Build GRCh38
rs140586410 Chr.18: 58517104-58517104 on Build GRCh38
rs1042694035 Chr.18: 58517150-58517150 on Build GRCh38
rs1049151623 Chr.18: 58517178-58517178 on Build GRCh38
rs761210364 Chr.18: 58517181-58517181 on Build GRCh38
rs756164408 Chr.18: 58517140-58517142 on Build GRCh38
rs779510607 Chr.18: 58517037-58517037 on Build GRCh38
rs765310079 Chr.18: 58516972-58516972 on Build GRCh38
rs750717625 Chr.18: 58517027-58517027 on Build GRCh38
rs777547519 Chr.18: 58517074-58517074 on Build GRCh38
rs762342746 Chr.18: 58517088-58517088 on Build GRCh38
rs201097473 Chr.18: 58517170-58517170 on Build GRCh38
rs34823643 Chr.18: 58517111-58517111 on Build GRCh38
rs1010319556 Chr.18: 58517146-58517146 on Build GRCh38
rs56067275 Chr.18: 58517057-58517057 on Build GRCh38
rs775644161 Chr.18: 58517077-58517077 on Build GRCh38
rs781021988 Chr.18: 58517142-58517142 on Build GRCh38
rs781603343 Chr.18: 58517063-58517063 on Build GRCh38
rs755791994 Chr.18: 58517033-58517033 on Build GRCh38
rs768775259 Chr.18: 58517079-58517079 on Build GRCh38
rs766701696 Chr.18: 58517182-58517182 on Build GRCh38
rs143318124 Chr.18: 58517068-58517068 on Build GRCh38
rs766759599 Chr.18: 58517094-58517094 on Build GRCh38
rs1041293229 Chr.18: 58517113-58517113 on Build GRCh38
rs368495288 Chr.18: 58517179-58517179 on Build GRCh38
rs563273481 Chr.18: 58517121-58517121 on Build GRCh38
rs751872060 Chr.18: 58517017-58517017 on Build GRCh38
rs764155757 Chr.18: 58516993-58516993 on Build GRCh38
rs530701876 Chr.18: 58517160-58517160 on Build GRCh38

View Full Product Details