Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32401606-32401865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL2
Entrez Gene ID
56244
Gene Name
butyrophilin like 2
Gene Aliases
BTL-II, BTN7, HSBLMHC1, SS2
UniGene
Hs.534471
Gene Chromosome Location
Chr.6: 32393339-32408879 on Build GRCh38
Transcripts
NM_001304561.1 XM_017011057.1 NR_136245.1

Gene Symbol
LOC101929163
Entrez Gene ID
101929163
Gene Name
uncharacterized LOC101929163
Gene Aliases
-
UniGene
Hs.57692
Gene Chromosome Location
Chr.6: 32255173-32407730 on Build GRCh38
Transcripts
NM_001304561.1 XM_017011057.1 NR_136245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4354453 c.720G>A p.Q240Q Chr.6: 32401795-32401795 on Build GRCh38
6351064 c.730+8C>T p.? Chr.6: 32401777-32401777 on Build GRCh38
6351063 c.730+8C>T p.? Chr.6: 32401777-32401777 on Build GRCh38
3625434 c.711G>A p.E237E Chr.6: 32401804-32401804 on Build GRCh38
3348717 c.720G>A p.Q240Q Chr.6: 32401795-32401795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117335692 Chr.6: 32401651-32401651 on Build GRCh38
rs954457768 Chr.6: 32401665-32401665 on Build GRCh38
rs920835547 Chr.6: 32401703-32401703 on Build GRCh38
rs140893144 Chr.6: 32401785-32401785 on Build GRCh38
rs779146636 Chr.6: 32401759-32401759 on Build GRCh38
rs117674475 Chr.6: 32401662-32401662 on Build GRCh38
rs767439942 Chr.6: 32401830-32401830 on Build GRCh38
rs555501721 Chr.6: 32401701-32401703 on Build GRCh38
rs117501081 Chr.6: 32401689-32401689 on Build GRCh38
rs759181827 Chr.6: 32401749-32401750 on Build GRCh38
rs772404653 Chr.6: 32401764-32401764 on Build GRCh38
rs117080666 Chr.6: 32401819-32401819 on Build GRCh38
rs117065092 Chr.6: 32401677-32401677 on Build GRCh38
rs538548316 Chr.6: 32401813-32401813 on Build GRCh38
rs760059055 Chr.6: 32401784-32401784 on Build GRCh38
rs148573746 Chr.6: 32401810-32401811 on Build GRCh38
rs1039609591 Chr.6: 32401792-32401792 on Build GRCh38
rs3763305 Chr.6: 32401711-32401711 on Build GRCh38
rs755057103 Chr.6: 32401752-32401752 on Build GRCh38
rs752142966 Chr.6: 32401819-32401821 on Build GRCh38
rs377488060 Chr.6: 32401816-32401817 on Build GRCh38
rs747450880 Chr.6: 32401774-32401774 on Build GRCh38
rs990248313 Chr.6: 32401708-32401708 on Build GRCh38
rs537877812 Chr.6: 32401762-32401762 on Build GRCh38
rs372954436 Chr.6: 32401820-32401820 on Build GRCh38
rs117084565 Chr.6: 32401740-32401740 on Build GRCh38
rs777986116 Chr.6: 32401772-32401772 on Build GRCh38
rs41399051 Chr.6: 32401824-32401824 on Build GRCh38
rs750606254 Chr.6: 32401776-32401776 on Build GRCh38
rs958967804 Chr.6: 32401705-32401705 on Build GRCh38
rs191295850 Chr.6: 32401817-32401817 on Build GRCh38
rs750415627 Chr.6: 32401750-32401750 on Build GRCh38
rs113258800 Chr.6: 32401828-32401828 on Build GRCh38
rs961755548 Chr.6: 32401697-32401697 on Build GRCh38
rs995951460 Chr.6: 32401658-32401658 on Build GRCh38
rs995109692 Chr.6: 32401825-32401825 on Build GRCh38
rs116191755 Chr.6: 32401777-32401777 on Build GRCh38

View Full Product Details