Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30924228-30924499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEDAG
Entrez Gene ID
84935
Gene Name
mesenteric estrogen dependent adipogenesis
Gene Aliases
AWMS3, C13orf33, MEDA-4, MEDA4, hAWMS3
UniGene
Hs.147880
Gene Chromosome Location
Chr.13: 30906175-30925577 on Build GRCh38
Transcripts
NM_032849.3 XM_017020801.1 NR_038287.1

Gene Symbol
TEX26-AS1
Entrez Gene ID
100507064
Gene Name
TEX26 antisense RNA 1
Gene Aliases
LINC00447
UniGene
Hs.654878
Gene Chromosome Location
Chr.13: 30882835-30932608 on Build GRCh38
Transcripts
NM_032849.3 XM_017020801.1 NR_038287.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5518012 c.804T>C p.V268V Chr.13: 30924327-30924327 on Build GRCh38
5012654 c.826A>G p.R276G Chr.13: 30924349-30924349 on Build GRCh38
2070876 c.803delT p.F269fs*11 Chr.13: 30924326-30924326 on Build GRCh38
6711037 c.836T>G p.L279R Chr.13: 30924359-30924359 on Build GRCh38
5620016 c.863C>T p.P288L Chr.13: 30924386-30924386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757561985 Chr.13: 30924328-30924328 on Build GRCh38
rs761558165 Chr.13: 30924277-30924277 on Build GRCh38
rs763531328 Chr.13: 30924394-30924394 on Build GRCh38
rs780202535 Chr.13: 30924470-30924470 on Build GRCh38
rs753349518 Chr.13: 30924295-30924295 on Build GRCh38
rs759087951 Chr.13: 30924384-30924384 on Build GRCh38
rs534073531 Chr.13: 30924270-30924270 on Build GRCh38
rs764835720 Chr.13: 30924386-30924386 on Build GRCh38
rs761196172 Chr.13: 30924270-30924291 on Build GRCh38
rs368825437 Chr.13: 30924310-30924310 on Build GRCh38
rs749266707 Chr.13: 30924478-30924478 on Build GRCh38
rs749956979 Chr.13: 30924323-30924324 on Build GRCh38
rs898213586 Chr.13: 30924473-30924473 on Build GRCh38
rs756193550 Chr.13: 30924334-30924334 on Build GRCh38
rs938936493 Chr.13: 30924351-30924351 on Build GRCh38
rs757686869 Chr.13: 30924441-30924441 on Build GRCh38
rs577992797 Chr.13: 30924253-30924253 on Build GRCh38
rs765956435 Chr.13: 30924293-30924293 on Build GRCh38
rs376363416 Chr.13: 30924292-30924292 on Build GRCh38
rs755425569 Chr.13: 30924418-30924418 on Build GRCh38
rs1050040158 Chr.13: 30924257-30924257 on Build GRCh38
rs566702242 Chr.13: 30924261-30924261 on Build GRCh38
rs767896228 Chr.13: 30924446-30924446 on Build GRCh38
rs61947585 Chr.13: 30924272-30924272 on Build GRCh38
rs914631778 Chr.13: 30924256-30924256 on Build GRCh38
rs759034703 Chr.13: 30924298-30924298 on Build GRCh38
rs992261307 Chr.13: 30924353-30924353 on Build GRCh38
rs751918546 Chr.13: 30924324-30924324 on Build GRCh38
rs747267893 Chr.13: 30924262-30924262 on Build GRCh38
rs781564848 Chr.13: 30924330-30924330 on Build GRCh38
rs758793268 Chr.13: 30924319-30924319 on Build GRCh38
rs771854199 Chr.13: 30924364-30924364 on Build GRCh38
rs773002689 Chr.13: 30924370-30924370 on Build GRCh38
rs765714090 Chr.13: 30924299-30924299 on Build GRCh38
rs764445175 Chr.13: 30924398-30924398 on Build GRCh38
rs776243503 Chr.13: 30924379-30924379 on Build GRCh38
rs764774458 Chr.13: 30924303-30924303 on Build GRCh38
rs751810162 Chr.13: 30924330-30924331 on Build GRCh38
rs779681405 Chr.13: 30924329-30924330 on Build GRCh38
rs201393057 Chr.13: 30924266-30924266 on Build GRCh38
rs750638670 Chr.13: 30924464-30924464 on Build GRCh38
rs146086897 Chr.13: 30924337-30924337 on Build GRCh38
rs770637457 Chr.13: 30924378-30924378 on Build GRCh38
rs758005318 Chr.13: 30924326-30924326 on Build GRCh38
rs774815996 Chr.13: 30924388-30924388 on Build GRCh38
rs146713994 Chr.13: 30924465-30924465 on Build GRCh38
rs748145597 Chr.13: 30924338-30924338 on Build GRCh38
rs745881119 Chr.13: 30924271-30924271 on Build GRCh38
rs745999526 Chr.13: 30924332-30924332 on Build GRCh38
rs760973126 Chr.13: 30924341-30924341 on Build GRCh38
rs577265236 Chr.13: 30924290-30924290 on Build GRCh38
rs771867109 Chr.13: 30924285-30924285 on Build GRCh38
rs746739528 Chr.13: 30924373-30924373 on Build GRCh38
rs373323371 Chr.13: 30924410-30924410 on Build GRCh38

View Full Product Details