Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29326393-29326664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACH1
Entrez Gene ID
571
Gene Name
BTB domain and CNC homolog 1
Gene Aliases
BACH-1, BTBD24
UniGene
Hs.154276
Gene Chromosome Location
Chr.21: 29298795-29361896 on Build GRCh38
Transcripts
NM_001186.3 NM_206866.2 NR_027655.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6609521 c.646G>A p.A216T Chr.21: 29326470-29326470 on Build GRCh38
309416 c.597A>G p.L199L Chr.21: 29326421-29326421 on Build GRCh38
1029785 c.683G>T p.R228I Chr.21: 29326507-29326507 on Build GRCh38
6559069 c.717C>T p.V239V Chr.21: 29326541-29326541 on Build GRCh38
7357226 c.729A>T p.E243D Chr.21: 29326553-29326553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545357951 Chr.21: 29326574-29326574 on Build GRCh38
rs374723065 Chr.21: 29326417-29326417 on Build GRCh38
rs757377063 Chr.21: 29326631-29326631 on Build GRCh38
rs1001859623 Chr.21: 29326493-29326493 on Build GRCh38
rs574088955 Chr.21: 29326496-29326496 on Build GRCh38
rs553355487 Chr.21: 29326543-29326543 on Build GRCh38
rs970456121 Chr.21: 29326474-29326474 on Build GRCh38
rs61735773 Chr.21: 29326451-29326451 on Build GRCh38
rs768118683 Chr.21: 29326463-29326463 on Build GRCh38
rs779145687 Chr.21: 29326537-29326537 on Build GRCh38
rs757462849 Chr.21: 29326521-29326521 on Build GRCh38
rs746048388 Chr.21: 29326633-29326633 on Build GRCh38
rs368476331 Chr.21: 29326542-29326542 on Build GRCh38
rs753098656 Chr.21: 29326481-29326481 on Build GRCh38
rs138981213 Chr.21: 29326452-29326452 on Build GRCh38
rs746856749 Chr.21: 29326547-29326547 on Build GRCh38
rs374802824 Chr.21: 29326626-29326626 on Build GRCh38
rs542608015 Chr.21: 29326513-29326513 on Build GRCh38
rs140102895 Chr.21: 29326425-29326425 on Build GRCh38
rs756562023 Chr.21: 29326627-29326627 on Build GRCh38
rs771966161 Chr.21: 29326423-29326423 on Build GRCh38
rs372369161 Chr.21: 29326628-29326628 on Build GRCh38
rs372118522 Chr.21: 29326549-29326549 on Build GRCh38
rs764495595 Chr.21: 29326611-29326611 on Build GRCh38
rs371081000 Chr.21: 29326442-29326442 on Build GRCh38
rs4817282 Chr.21: 29326634-29326634 on Build GRCh38
rs762721028 Chr.21: 29326619-29326619 on Build GRCh38
rs147096876 Chr.21: 29326562-29326562 on Build GRCh38
rs536766184 Chr.21: 29326453-29326453 on Build GRCh38
rs200303423 Chr.21: 29326460-29326460 on Build GRCh38
rs34811108 Chr.21: 29326553-29326553 on Build GRCh38
rs745638584 Chr.21: 29326539-29326539 on Build GRCh38
rs764957109 Chr.21: 29326572-29326572 on Build GRCh38
rs61735774 Chr.21: 29326632-29326632 on Build GRCh38
rs147328273 Chr.21: 29326427-29326427 on Build GRCh38
rs767704566 Chr.21: 29326624-29326624 on Build GRCh38
rs201844220 Chr.21: 29326506-29326506 on Build GRCh38
rs957755223 Chr.21: 29326497-29326497 on Build GRCh38
rs375713948 Chr.21: 29326456-29326456 on Build GRCh38
rs763472575 Chr.21: 29326437-29326437 on Build GRCh38

View Full Product Details