Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18021672-18021939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATPAF2
Entrez Gene ID
91647
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 2
Gene Aliases
ATP12, ATP12p, LP3663, MC5DN1
UniGene
Hs.528889
Gene Chromosome Location
Chr.17: 18015059-18039169 on Build GRCh38
Transcripts
NM_145691.3 XM_005256848.3 XM_011524065.1 XM_017025302.1 XM_017025303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3958075 c.607G>A p.A203T Chr.17: 18021754-18021754 on Build GRCh38
976177 c.507C>T p.Y169Y Chr.17: 18021854-18021854 on Build GRCh38
5513273 c.589G>A p.A197T Chr.17: 18021772-18021772 on Build GRCh38
4498578 c.522C>T p.S174S Chr.17: 18021839-18021839 on Build GRCh38
7351537 c.504-1G>C p.? Chr.17: 18021858-18021858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750424854 Chr.17: 18021865-18021865 on Build GRCh38
rs746996386 Chr.17: 18021871-18021871 on Build GRCh38
rs756519716 Chr.17: 18021764-18021764 on Build GRCh38
rs762338846 Chr.17: 18021905-18021905 on Build GRCh38
rs771868422 Chr.17: 18021791-18021791 on Build GRCh38
rs760581066 Chr.17: 18021723-18021723 on Build GRCh38
rs201900136 Chr.17: 18021873-18021873 on Build GRCh38
rs539903484 Chr.17: 18021725-18021725 on Build GRCh38
rs761399673 Chr.17: 18021727-18021727 on Build GRCh38
rs367939316 Chr.17: 18021853-18021853 on Build GRCh38
rs778802460 Chr.17: 18021784-18021784 on Build GRCh38
rs150185337 Chr.17: 18021815-18021815 on Build GRCh38
rs775360731 Chr.17: 18021718-18021718 on Build GRCh38
rs779229175 Chr.17: 18021859-18021859 on Build GRCh38
rs758558880 Chr.17: 18021867-18021867 on Build GRCh38
rs141827488 Chr.17: 18021796-18021796 on Build GRCh38
rs762546188 Chr.17: 18021826-18021826 on Build GRCh38
rs17853022 Chr.17: 18021839-18021839 on Build GRCh38
rs780166761 Chr.17: 18021795-18021795 on Build GRCh38
rs201082226 Chr.17: 18021881-18021881 on Build GRCh38
rs777940883 Chr.17: 18021765-18021765 on Build GRCh38
rs774341906 Chr.17: 18021695-18021695 on Build GRCh38
rs968149971 Chr.17: 18021876-18021876 on Build GRCh38
rs772965371 Chr.17: 18021882-18021882 on Build GRCh38
rs979498082 Chr.17: 18021878-18021878 on Build GRCh38
rs769704888 Chr.17: 18021727-18021730 on Build GRCh38
rs757347605 Chr.17: 18021780-18021780 on Build GRCh38
rs757357549 Chr.17: 18021854-18021854 on Build GRCh38
rs369318001 Chr.17: 18021785-18021785 on Build GRCh38
rs62073570 Chr.17: 18021850-18021850 on Build GRCh38
rs769489117 Chr.17: 18021816-18021816 on Build GRCh38
rs772061430 Chr.17: 18021716-18021716 on Build GRCh38
rs770092762 Chr.17: 18021708-18021708 on Build GRCh38
rs1008083351 Chr.17: 18021857-18021857 on Build GRCh38
rs200680173 Chr.17: 18021715-18021715 on Build GRCh38
rs757840763 Chr.17: 18021747-18021747 on Build GRCh38
rs781064154 Chr.17: 18021877-18021877 on Build GRCh38
rs767887072 Chr.17: 18021829-18021829 on Build GRCh38
rs749960290 Chr.17: 18021739-18021739 on Build GRCh38
rs745851759 Chr.17: 18021698-18021698 on Build GRCh38
rs776406701 Chr.17: 18021813-18021813 on Build GRCh38
rs942295101 Chr.17: 18021730-18021730 on Build GRCh38
rs913902683 Chr.17: 18021875-18021875 on Build GRCh38
rs757002424 Chr.17: 18021851-18021851 on Build GRCh38
rs553417858 Chr.17: 18021742-18021742 on Build GRCh38
rs375133434 Chr.17: 18021741-18021741 on Build GRCh38
rs749579388 Chr.17: 18021772-18021772 on Build GRCh38
rs764708359 Chr.17: 18021738-18021738 on Build GRCh38
rs113782839 Chr.17: 18021908-18021908 on Build GRCh38
rs772554573 Chr.17: 18021823-18021823 on Build GRCh38
rs753242638 Chr.17: 18021831-18021831 on Build GRCh38
rs748004339 Chr.17: 18021879-18021879 on Build GRCh38

View Full Product Details