Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18907069-18907296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPE
Entrez Gene ID
11316
Gene Name
coatomer protein complex subunit epsilon
Gene Aliases
epsilon-COP
UniGene
Hs.10326
Gene Chromosome Location
Chr.19: 18899514-18919464 on Build GRCh38
Transcripts
NM_007263.3 NM_199442.1 NM_199444.1 XM_011527655.1 XM_011527656.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3770233 c.306C>T p.A102A Chr.19: 18907097-18907097 on Build GRCh38
5806199 c.304G>C p.A102P Chr.19: 18907099-18907099 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374423155 Chr.19: 18907125-18907125 on Build GRCh38
rs147402162 Chr.19: 18907245-18907245 on Build GRCh38
rs753354317 Chr.19: 18907141-18907141 on Build GRCh38
rs750309430 Chr.19: 18907134-18907134 on Build GRCh38
rs145639224 Chr.19: 18907090-18907090 on Build GRCh38
rs986080500 Chr.19: 18907240-18907240 on Build GRCh38
rs747843335 Chr.19: 18907173-18907174 on Build GRCh38
rs776760562 Chr.19: 18907103-18907103 on Build GRCh38
rs10164368 Chr.19: 18907191-18907191 on Build GRCh38
rs917999271 Chr.19: 18907171-18907171 on Build GRCh38
rs759184749 Chr.19: 18907244-18907244 on Build GRCh38
rs913305850 Chr.19: 18907262-18907262 on Build GRCh38
rs147731610 Chr.19: 18907097-18907097 on Build GRCh38
rs760823251 Chr.19: 18907096-18907096 on Build GRCh38
rs535193620 Chr.19: 18907228-18907228 on Build GRCh38
rs371709319 Chr.19: 18907137-18907137 on Build GRCh38
rs139812875 Chr.19: 18907252-18907252 on Build GRCh38
rs376032669 Chr.19: 18907158-18907158 on Build GRCh38
rs769297059 Chr.19: 18907268-18907268 on Build GRCh38
rs755811596 Chr.19: 18907136-18907136 on Build GRCh38
rs759584150 Chr.19: 18907106-18907106 on Build GRCh38
rs961539376 Chr.19: 18907157-18907157 on Build GRCh38
rs776443530 Chr.19: 18907232-18907232 on Build GRCh38
rs980009515 Chr.19: 18907102-18907102 on Build GRCh38
rs767352017 Chr.19: 18907133-18907133 on Build GRCh38
rs773583293 Chr.19: 18907092-18907092 on Build GRCh38
rs938528864 Chr.19: 18907140-18907140 on Build GRCh38
rs373983247 Chr.19: 18907087-18907087 on Build GRCh38
rs754401855 Chr.19: 18907144-18907144 on Build GRCh38
rs760167601 Chr.19: 18907120-18907121 on Build GRCh38
rs199509192 Chr.19: 18907131-18907131 on Build GRCh38

View Full Product Details