Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8060089-8060314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3AR1
Entrez Gene ID
719
Gene Name
complement component 3a receptor 1
Gene Aliases
AZ3B, C3AR, HNFAG09
UniGene
Hs.504656
Gene Chromosome Location
Chr.12: 8058323-8066359 on Build GRCh38
Transcripts
NM_001326475.1 NM_001326477.1 NM_004054.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
695639 c.89T>A p.L30H Chr.12: 8060097-8060097 on Build GRCh38
4045120 c.5C>T p.A2V Chr.12: 8060181-8060181 on Build GRCh38
4550389 c.4G>A p.A2T Chr.12: 8060182-8060182 on Build GRCh38
6203886 c.47A>T p.Q16L Chr.12: 8060139-8060139 on Build GRCh38
7280766 c.33T>C p.T11T Chr.12: 8060153-8060153 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs931861654 Chr.12: 8060114-8060114 on Build GRCh38
rs151033908 Chr.12: 8060149-8060149 on Build GRCh38
rs759779343 Chr.12: 8060234-8060234 on Build GRCh38
rs758721088 Chr.12: 8060249-8060249 on Build GRCh38
rs868285987 Chr.12: 8060178-8060178 on Build GRCh38
rs1022158562 Chr.12: 8060289-8060289 on Build GRCh38
rs556397063 Chr.12: 8060252-8060252 on Build GRCh38
rs763894948 Chr.12: 8060131-8060131 on Build GRCh38
rs907480919 Chr.12: 8060276-8060276 on Build GRCh38
rs1048590619 Chr.12: 8060141-8060141 on Build GRCh38
rs774625175 Chr.12: 8060232-8060232 on Build GRCh38
rs7968865 Chr.12: 8060286-8060286 on Build GRCh38
rs757061171 Chr.12: 8060148-8060148 on Build GRCh38
rs140906071 Chr.12: 8060180-8060180 on Build GRCh38
rs749671193 Chr.12: 8060197-8060197 on Build GRCh38
rs11567804 Chr.12: 8060196-8060196 on Build GRCh38
rs771275173 Chr.12: 8060218-8060218 on Build GRCh38
rs767599737 Chr.12: 8060119-8060119 on Build GRCh38
rs900822877 Chr.12: 8060206-8060206 on Build GRCh38
rs779604521 Chr.12: 8060193-8060193 on Build GRCh38
rs1010264891 Chr.12: 8060227-8060227 on Build GRCh38
rs752624182 Chr.12: 8060124-8060124 on Build GRCh38
rs772132268 Chr.12: 8060235-8060235 on Build GRCh38
rs369213108 Chr.12: 8060152-8060152 on Build GRCh38
rs761311473 Chr.12: 8060220-8060224 on Build GRCh38
rs770364981 Chr.12: 8060186-8060186 on Build GRCh38
rs953853770 Chr.12: 8060275-8060275 on Build GRCh38
rs753689444 Chr.12: 8060146-8060146 on Build GRCh38
rs779595963 Chr.12: 8060183-8060183 on Build GRCh38
rs756095569 Chr.12: 8060126-8060126 on Build GRCh38
rs371493019 Chr.12: 8060181-8060181 on Build GRCh38

View Full Product Details