Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122742059-122742310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBS12
Entrez Gene ID
166379
Gene Name
Bardet-Biedl syndrome 12
Gene Aliases
C4orf24
UniGene
Hs.400698
Gene Chromosome Location
Chr.4: 122700437-122744943 on Build GRCh38
Transcripts
NM_001178007.1 NM_152618.2 XM_011531680.2 XM_017007831.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7357326 c.316G>T p.G106C Chr.4: 122742208-122742208 on Build GRCh38
5045887 c.214G>T p.E72* Chr.4: 122742106-122742106 on Build GRCh38
7107463 c.285G>C p.W95C Chr.4: 122742177-122742177 on Build GRCh38
73812 c.370A>T p.S124C Chr.4: 122742262-122742262 on Build GRCh38
6610128 c.386C>A p.S129Y Chr.4: 122742278-122742278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201693990 Chr.4: 122742154-122742154 on Build GRCh38
rs375439566 Chr.4: 122742274-122742274 on Build GRCh38
rs56320045 Chr.4: 122742263-122742263 on Build GRCh38
rs778171614 Chr.4: 122742103-122742103 on Build GRCh38
rs1052836009 Chr.4: 122742093-122742093 on Build GRCh38
rs759091753 Chr.4: 122742208-122742208 on Build GRCh38
rs755940610 Chr.4: 122742097-122742097 on Build GRCh38
rs758963348 Chr.4: 122742121-122742121 on Build GRCh38
rs767601166 Chr.4: 122742228-122742228 on Build GRCh38
rs768973035 Chr.4: 122742133-122742133 on Build GRCh38
rs763730747 Chr.4: 122742261-122742261 on Build GRCh38
rs770609788 Chr.4: 122742182-122742182 on Build GRCh38
rs934909698 Chr.4: 122742268-122742268 on Build GRCh38
rs309369 Chr.4: 122742270-122742270 on Build GRCh38
rs144179606 Chr.4: 122742236-122742236 on Build GRCh38
rs772942987 Chr.4: 122742135-122742135 on Build GRCh38
rs973844350 Chr.4: 122742166-122742166 on Build GRCh38
rs767546051 Chr.4: 122742092-122742092 on Build GRCh38
rs587777801 Chr.4: 122742227-122742229 on Build GRCh38
rs752586913 Chr.4: 122742094-122742094 on Build GRCh38
rs773788808 Chr.4: 122742188-122742188 on Build GRCh38
rs180723721 Chr.4: 122742223-122742223 on Build GRCh38
rs745957664 Chr.4: 122742119-122742119 on Build GRCh38
rs778792181 Chr.4: 122742114-122742114 on Build GRCh38
rs915677674 Chr.4: 122742203-122742203 on Build GRCh38
rs754104512 Chr.4: 122742269-122742269 on Build GRCh38
rs373273487 Chr.4: 122742129-122742129 on Build GRCh38
rs77731085 Chr.4: 122742247-122742247 on Build GRCh38
rs143960329 Chr.4: 122742104-122742104 on Build GRCh38
rs746271266 Chr.4: 122742155-122742155 on Build GRCh38
rs151344630 Chr.4: 122742215-122742215 on Build GRCh38
rs528175399 Chr.4: 122742226-122742226 on Build GRCh38
rs907426686 Chr.4: 122742258-122742258 on Build GRCh38

View Full Product Details