Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23684518-23684781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM221A
Entrez Gene ID
340277
Gene Name
family with sequence similarity 221 member A
Gene Aliases
C7orf46
UniGene
Hs.592178
Gene Chromosome Location
Chr.7: 23680114-23703249 on Build GRCh38
Transcripts
NM_001127364.2 NM_001127365.2 NM_001300932.1 NM_199136.4 XM_011515369.2 XM_011515370.2 XM_011515371.2 XM_011515372.2 XM_017012134.1 XM_017012135.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5733092 c.216G>C p.E72D Chr.7: 23684649-23684649 on Build GRCh38
3026242 c.176G>T p.R59L Chr.7: 23684609-23684609 on Build GRCh38
3026241 c.174G>A p.W58* Chr.7: 23684607-23684607 on Build GRCh38
3880289 c.155A>C p.N52T Chr.7: 23684588-23684588 on Build GRCh38
4521198 c.109G>A p.E37K Chr.7: 23684542-23684542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749626297 Chr.7: 23684613-23684613 on Build GRCh38
rs760732004 Chr.7: 23684628-23684628 on Build GRCh38
rs751046668 Chr.7: 23684676-23684676 on Build GRCh38
rs765157798 Chr.7: 23684652-23684652 on Build GRCh38
rs540797833 Chr.7: 23684547-23684547 on Build GRCh38
rs779349956 Chr.7: 23684617-23684617 on Build GRCh38
rs368760611 Chr.7: 23684675-23684675 on Build GRCh38
rs752705562 Chr.7: 23684573-23684573 on Build GRCh38
rs1013481338 Chr.7: 23684639-23684639 on Build GRCh38
rs376004928 Chr.7: 23684645-23684645 on Build GRCh38
rs375175216 Chr.7: 23684564-23684564 on Build GRCh38
rs772627182 Chr.7: 23684713-23684713 on Build GRCh38
rs139023569 Chr.7: 23684578-23684578 on Build GRCh38
rs759444304 Chr.7: 23684558-23684558 on Build GRCh38
rs371705612 Chr.7: 23684682-23684682 on Build GRCh38
rs1002438833 Chr.7: 23684589-23684589 on Build GRCh38
rs766567064 Chr.7: 23684548-23684548 on Build GRCh38
rs113158506 Chr.7: 23684618-23684618 on Build GRCh38
rs777042503 Chr.7: 23684644-23684644 on Build GRCh38
rs550013218 Chr.7: 23684723-23684723 on Build GRCh38
rs548646278 Chr.7: 23684608-23684608 on Build GRCh38
rs7791123 Chr.7: 23684745-23684745 on Build GRCh38
rs754072871 Chr.7: 23684590-23684591 on Build GRCh38
rs751118991 Chr.7: 23684582-23684582 on Build GRCh38
rs368536274 Chr.7: 23684614-23684614 on Build GRCh38
rs372615304 Chr.7: 23684679-23684679 on Build GRCh38
rs771985246 Chr.7: 23684619-23684619 on Build GRCh38
rs753683025 Chr.7: 23684557-23684557 on Build GRCh38
rs867175543 Chr.7: 23684585-23684585 on Build GRCh38
rs771153860 Chr.7: 23684631-23684631 on Build GRCh38
rs779260475 Chr.7: 23684688-23684688 on Build GRCh38
rs777343625 Chr.7: 23684579-23684579 on Build GRCh38
rs756762053 Chr.7: 23684677-23684677 on Build GRCh38
rs780771415 Chr.7: 23684678-23684678 on Build GRCh38
rs780859206 Chr.7: 23684609-23684609 on Build GRCh38
rs773327664 Chr.7: 23684626-23684626 on Build GRCh38
rs752617705 Chr.7: 23684663-23684663 on Build GRCh38
rs762965869 Chr.7: 23684669-23684669 on Build GRCh38
rs190417461 Chr.7: 23684694-23684694 on Build GRCh38

View Full Product Details