Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 35659091-35659346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOL6
Entrez Gene ID
80830
Gene Name
apolipoprotein L6
Gene Aliases
APOL-VI, APOLVI
UniGene
Hs.257352
Gene Chromosome Location
Chr.22: 35648317-35668409 on Build GRCh38
Transcripts
NM_030641.3 XM_011530392.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3729772 c.683C>T p.A228V Chr.22: 35659247-35659247 on Build GRCh38
4752760 c.676A>C p.T226P Chr.22: 35659240-35659240 on Build GRCh38
1714326 c.754C>T p.L252F Chr.22: 35659318-35659318 on Build GRCh38
6095070 c.748G>T p.A250S Chr.22: 35659312-35659312 on Build GRCh38
4888978 c.763G>A p.E255K Chr.22: 35659327-35659327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201496034 Chr.22: 35659171-35659171 on Build GRCh38
rs930875786 Chr.22: 35659318-35659318 on Build GRCh38
rs140728642 Chr.22: 35659185-35659185 on Build GRCh38
rs752702500 Chr.22: 35659248-35659248 on Build GRCh38
rs758262301 Chr.22: 35659202-35659202 on Build GRCh38
rs540748972 Chr.22: 35659253-35659253 on Build GRCh38
rs748325179 Chr.22: 35659309-35659312 on Build GRCh38
rs774975141 Chr.22: 35659212-35659212 on Build GRCh38
rs573758358 Chr.22: 35659287-35659287 on Build GRCh38
rs754753929 Chr.22: 35659116-35659116 on Build GRCh38
rs762282595 Chr.22: 35659159-35659159 on Build GRCh38
rs374093921 Chr.22: 35659233-35659233 on Build GRCh38
rs772267988 Chr.22: 35659314-35659359 on Build GRCh38
rs746692910 Chr.22: 35659204-35659204 on Build GRCh38
rs201669216 Chr.22: 35659234-35659234 on Build GRCh38
rs145863080 Chr.22: 35659210-35659210 on Build GRCh38
rs770447719 Chr.22: 35659132-35659132 on Build GRCh38
rs192850920 Chr.22: 35659121-35659121 on Build GRCh38
rs200500746 Chr.22: 35659208-35659208 on Build GRCh38
rs752403368 Chr.22: 35659201-35659201 on Build GRCh38
rs772269044 Chr.22: 35659283-35659283 on Build GRCh38
rs765286120 Chr.22: 35659192-35659192 on Build GRCh38
rs146609959 Chr.22: 35659266-35659266 on Build GRCh38
rs560610184 Chr.22: 35659265-35659265 on Build GRCh38
rs751152444 Chr.22: 35659264-35659264 on Build GRCh38
rs771248197 Chr.22: 35659304-35659304 on Build GRCh38
rs775935414 Chr.22: 35659141-35659141 on Build GRCh38
rs141435261 Chr.22: 35659288-35659288 on Build GRCh38
rs777040698 Chr.22: 35659316-35659316 on Build GRCh38
rs746500379 Chr.22: 35659123-35659123 on Build GRCh38
rs558213377 Chr.22: 35659220-35659220 on Build GRCh38
rs763953185 Chr.22: 35659255-35659255 on Build GRCh38
rs773507616 Chr.22: 35659172-35659172 on Build GRCh38
rs749797800 Chr.22: 35659142-35659142 on Build GRCh38
rs745436533 Chr.22: 35659209-35659209 on Build GRCh38
rs200170835 Chr.22: 35659150-35659150 on Build GRCh38
rs996121781 Chr.22: 35659251-35659251 on Build GRCh38
rs749494425 Chr.22: 35659161-35659162 on Build GRCh38
rs190149914 Chr.22: 35659117-35659117 on Build GRCh38
rs774562457 Chr.22: 35659151-35659151 on Build GRCh38
rs772520677 Chr.22: 35659219-35659219 on Build GRCh38
rs200966307 Chr.22: 35659176-35659176 on Build GRCh38
rs770840573 Chr.22: 35659207-35659207 on Build GRCh38
rs777240326 Chr.22: 35659203-35659203 on Build GRCh38
rs779250565 Chr.22: 35659272-35659272 on Build GRCh38
rs953824475 Chr.22: 35659133-35659133 on Build GRCh38
rs143314363 Chr.22: 35659273-35659273 on Build GRCh38
rs575853273 Chr.22: 35659206-35659206 on Build GRCh38
rs778861738 Chr.22: 35659221-35659222 on Build GRCh38
rs377394530 Chr.22: 35659178-35659178 on Build GRCh38
rs149179370 Chr.22: 35659267-35659267 on Build GRCh38
rs755169983 Chr.22: 35659167-35659168 on Build GRCh38
rs1044608188 Chr.22: 35659156-35659156 on Build GRCh38
rs576893978 Chr.22: 35659237-35659237 on Build GRCh38
rs148322582 Chr.22: 35659278-35659278 on Build GRCh38
rs760800006 Chr.22: 35659173-35659173 on Build GRCh38
rs369107640 Chr.22: 35659120-35659120 on Build GRCh38
rs760992573 Chr.22: 35659232-35659232 on Build GRCh38
rs759731291 Chr.22: 35659242-35659242 on Build GRCh38
rs1019237230 Chr.22: 35659285-35659285 on Build GRCh38
rs200520869 Chr.22: 35659247-35659247 on Build GRCh38
rs370175163 Chr.22: 35659254-35659254 on Build GRCh38
rs748819725 Chr.22: 35659180-35659180 on Build GRCh38
rs200146802 Chr.22: 35659218-35659218 on Build GRCh38
rs1044662290 Chr.22: 35659300-35659300 on Build GRCh38
rs753772999 Chr.22: 35659179-35659179 on Build GRCh38

View Full Product Details