Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186390670-186390895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf27
Entrez Gene ID
54953
Gene Name
chromosome 1 open reading frame 27
Gene Aliases
ODR4, TTG1, odr-4
UniGene
Hs.371210
Gene Chromosome Location
Chr.1: 186375758-186421375 on Build GRCh38
Transcripts
NM_001164245.1 NM_001164246.1 NM_017847.5 XM_011509660.1 XM_011509661.1 XM_017001543.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3369471 c.507A>G p.Q169Q Chr.1: 186390743-186390743 on Build GRCh38
1337223 c.531T>G p.L177L Chr.1: 186390767-186390767 on Build GRCh38
5622530 c.503A>T p.Y168F Chr.1: 186390739-186390739 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs139232938 Chr.1: 186390736-186390736 on Build GRCh38
rs773519415 Chr.1: 186390798-186390798 on Build GRCh38
rs769220502 Chr.1: 186390782-186390782 on Build GRCh38
rs992284252 Chr.1: 186390754-186390754 on Build GRCh38
rs750592224 Chr.1: 186390731-186390731 on Build GRCh38
rs752364848 Chr.1: 186390832-186390832 on Build GRCh38
rs765089806 Chr.1: 186390810-186390810 on Build GRCh38
rs764896739 Chr.1: 186390734-186390734 on Build GRCh38
rs187494832 Chr.1: 186390792-186390792 on Build GRCh38
rs371831284 Chr.1: 186390738-186390738 on Build GRCh38
rs781039619 Chr.1: 186390761-186390761 on Build GRCh38
rs201865055 Chr.1: 186390839-186390839 on Build GRCh38
rs759419943 Chr.1: 186390733-186390733 on Build GRCh38
rs751180543 Chr.1: 186390757-186390757 on Build GRCh38
rs376120716 Chr.1: 186390758-186390758 on Build GRCh38
rs772111453 Chr.1: 186390797-186390797 on Build GRCh38
rs776929590 Chr.1: 186390862-186390862 on Build GRCh38
rs766848929 Chr.1: 186390866-186390867 on Build GRCh38
rs368373111 Chr.1: 186390780-186390780 on Build GRCh38
rs780226436 Chr.1: 186390778-186390778 on Build GRCh38
rs774829162 Chr.1: 186390789-186390789 on Build GRCh38
rs777423436 Chr.1: 186390743-186390743 on Build GRCh38
rs745478585 Chr.1: 186390762-186390762 on Build GRCh38
rs919873302 Chr.1: 186390808-186390808 on Build GRCh38
rs375472243 Chr.1: 186390809-186390809 on Build GRCh38
rs761129676 Chr.1: 186390786-186390786 on Build GRCh38
rs770217455 Chr.1: 186390768-186390768 on Build GRCh38

View Full Product Details