Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 107319127-107319376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD11
Entrez Gene ID
121551
Gene Name
BTB domain containing 11
Gene Aliases
ABTB2B
UniGene
Hs.271272
Gene Chromosome Location
Chr.12: 107317168-107659642 on Build GRCh38
Transcripts
NM_001018072.1 XM_017018807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6248242 c.194G>T p.S65I Chr.12: 107319134-107319134 on Build GRCh38
4038315 c.205G>A p.V69I Chr.12: 107319145-107319145 on Build GRCh38
3356277 c.236G>T p.G79V Chr.12: 107319176-107319176 on Build GRCh38
5739780 c.248C>T p.A83V Chr.12: 107319188-107319188 on Build GRCh38
5763592 c.275C>T p.P92L Chr.12: 107319215-107319215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759054870 Chr.12: 107319243-107319243 on Build GRCh38
rs766089523 Chr.12: 107319226-107319226 on Build GRCh38
rs1033386964 Chr.12: 107319269-107319269 on Build GRCh38
rs753593336 Chr.12: 107319296-107319296 on Build GRCh38
rs751276608 Chr.12: 107319242-107319242 on Build GRCh38
rs752213419 Chr.12: 107319255-107319255 on Build GRCh38
rs767056116 Chr.12: 107319246-107319246 on Build GRCh38
rs770303401 Chr.12: 107319352-107319352 on Build GRCh38
rs755794696 Chr.12: 107319170-107319170 on Build GRCh38
rs919821876 Chr.12: 107319258-107319258 on Build GRCh38
rs973941049 Chr.12: 107319257-107319257 on Build GRCh38
rs763450951 Chr.12: 107319225-107319225 on Build GRCh38
rs917887068 Chr.12: 107319287-107319287 on Build GRCh38
rs777615818 Chr.12: 107319284-107319284 on Build GRCh38
rs755224319 Chr.12: 107319318-107319318 on Build GRCh38
rs749089539 Chr.12: 107319187-107319187 on Build GRCh38
rs773623357 Chr.12: 107319358-107319358 on Build GRCh38
rs373447496 Chr.12: 107319204-107319204 on Build GRCh38
rs952498058 Chr.12: 107319279-107319279 on Build GRCh38
rs747043614 Chr.12: 107319199-107319199 on Build GRCh38
rs556601692 Chr.12: 107319308-107319308 on Build GRCh38
rs781317817 Chr.12: 107319323-107319323 on Build GRCh38
rs74847588 Chr.12: 107319357-107319357 on Build GRCh38
rs747237962 Chr.12: 107319313-107319313 on Build GRCh38
rs377707357 Chr.12: 107319310-107319310 on Build GRCh38
rs937939485 Chr.12: 107319264-107319264 on Build GRCh38
rs748224177 Chr.12: 107319342-107319342 on Build GRCh38
rs772254650 Chr.12: 107319190-107319190 on Build GRCh38
rs183626910 Chr.12: 107319206-107319206 on Build GRCh38
rs780289076 Chr.12: 107319197-107319197 on Build GRCh38
rs773601656 Chr.12: 107319215-107319215 on Build GRCh38
rs983738669 Chr.12: 107319280-107319280 on Build GRCh38
rs986525193 Chr.12: 107319210-107319210 on Build GRCh38
rs768736328 Chr.12: 107319200-107319200 on Build GRCh38
rs777614203 Chr.12: 107319174-107319174 on Build GRCh38
rs776937868 Chr.12: 107319203-107319203 on Build GRCh38
rs540664436 Chr.12: 107319282-107319282 on Build GRCh38
rs1024906307 Chr.12: 107319231-107319231 on Build GRCh38

View Full Product Details