Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11354497-11354770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC159
Entrez Gene ID
126075
Gene Name
coiled-coil domain containing 159
Gene Aliases
-
UniGene
Hs.631636
Gene Chromosome Location
Chr.19: 11346340-11354951 on Build GRCh38
Transcripts
NM_001080503.2 XM_006722643.1 XM_011527678.2 XM_017026255.1 XM_017026256.1 XM_017026257.1 XM_017026258.1 XM_017026259.1 XM_017026260.1 XM_017026261.1 XM_017026262.1 XM_017026263.1 XM_017026264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1611538 c.1141A>T p.S381C Chr.19: 11354603-11354603 on Build GRCh38
990876 c.875G>A p.R292H Chr.19: 11354682-11354682 on Build GRCh38
5105020 c.828C>T p.S276S Chr.19: 11354635-11354635 on Build GRCh38
4989023 c.788A>T p.Q263L Chr.19: 11354595-11354595 on Build GRCh38
5775220 c.837C>G p.D279E Chr.19: 11354644-11354644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761062048 Chr.19: 11354715-11354715 on Build GRCh38
rs779225202 Chr.19: 11354682-11354682 on Build GRCh38
rs200875587 Chr.19: 11354719-11354719 on Build GRCh38
rs367946746 Chr.19: 11354605-11354605 on Build GRCh38
rs534702160 Chr.19: 11354589-11354591 on Build GRCh38
rs753535231 Chr.19: 11354538-11354538 on Build GRCh38
rs555253734 Chr.19: 11354525-11354525 on Build GRCh38
rs939415708 Chr.19: 11354747-11354747 on Build GRCh38
rs539681039 Chr.19: 11354594-11354611 on Build GRCh38
rs775535549 Chr.19: 11354636-11354636 on Build GRCh38
rs763917147 Chr.19: 11354714-11354715 on Build GRCh38
rs778943587 Chr.19: 11354631-11354631 on Build GRCh38
rs761437936 Chr.19: 11354544-11354544 on Build GRCh38
rs745696770 Chr.19: 11354632-11354632 on Build GRCh38
rs751602470 Chr.19: 11354715-11354715 on Build GRCh38
rs764968684 Chr.19: 11354654-11354654 on Build GRCh38
rs757235288 Chr.19: 11354726-11354726 on Build GRCh38
rs372176819 Chr.19: 11354677-11354677 on Build GRCh38
rs374274390 Chr.19: 11354635-11354635 on Build GRCh38
rs895426900 Chr.19: 11354610-11354610 on Build GRCh38
rs753226798 Chr.19: 11354602-11354602 on Build GRCh38
rs758285528 Chr.19: 11354705-11354705 on Build GRCh38
rs956433655 Chr.19: 11354739-11354739 on Build GRCh38
rs754732185 Chr.19: 11354735-11354735 on Build GRCh38
rs769405320 Chr.19: 11354750-11354750 on Build GRCh38
rs199507754 Chr.19: 11354696-11354696 on Build GRCh38
rs202020845 Chr.19: 11354680-11354680 on Build GRCh38
rs764628322 Chr.19: 11354679-11354679 on Build GRCh38
rs368424619 Chr.19: 11354720-11354720 on Build GRCh38
rs759987646 Chr.19: 11354660-11354660 on Build GRCh38
rs999658719 Chr.19: 11354666-11354666 on Build GRCh38
rs781149615 Chr.19: 11354736-11354736 on Build GRCh38
rs776002572 Chr.19: 11354709-11354709 on Build GRCh38
rs748980169 Chr.19: 11354721-11354721 on Build GRCh38
rs17001383 Chr.19: 11354606-11354606 on Build GRCh38
rs756006438 Chr.19: 11354684-11354684 on Build GRCh38
rs6887 Chr.19: 11354640-11354640 on Build GRCh38
rs375009614 Chr.19: 11354716-11354716 on Build GRCh38
rs374751855 Chr.19: 11354695-11354695 on Build GRCh38
rs772703372 Chr.19: 11354656-11354656 on Build GRCh38
rs187734834 Chr.19: 11354592-11354592 on Build GRCh38
rs751971157 Chr.19: 11354564-11354564 on Build GRCh38
rs762502253 Chr.19: 11354700-11354700 on Build GRCh38
rs771282974 Chr.19: 11354620-11354620 on Build GRCh38
rs746923821 Chr.19: 11354637-11354637 on Build GRCh38
rs767992542 Chr.19: 11354708-11354708 on Build GRCh38
rs776111294 Chr.19: 11354641-11354641 on Build GRCh38
rs371302789 Chr.19: 11354612-11354612 on Build GRCh38
rs755252779 Chr.19: 11354575-11354575 on Build GRCh38
rs758812671 Chr.19: 11354589-11354611 on Build GRCh38
rs757233994 Chr.19: 11354681-11354681 on Build GRCh38
rs201149711 Chr.19: 11354697-11354697 on Build GRCh38
rs752349077 Chr.19: 11354704-11354706 on Build GRCh38
rs987176932 Chr.19: 11354692-11354692 on Build GRCh38
rs763560777 Chr.19: 11354534-11354534 on Build GRCh38
rs537668849 Chr.19: 11354617-11354617 on Build GRCh38
rs748015867 Chr.19: 11354748-11354748 on Build GRCh38
rs769158442 Chr.19: 11354699-11354699 on Build GRCh38
rs767753487 Chr.19: 11354676-11354676 on Build GRCh38
rs764733230 Chr.19: 11354545-11354545 on Build GRCh38

View Full Product Details